Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43269267-43269496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG9
Entrez Gene ID
5678
Gene Name
pregnancy specific beta-1-glycoprotein 9
Gene Aliases
PS34, PSBG-11, PSBG-9, PSG11, PSGII
UniGene
Hs.502092
Gene Chromosome Location
Chr.19: 43253282-43269563 on Build GRCh38
Transcripts
NM_001301707.1 NM_001301708.1 NM_001301709.1 NM_002784.4 XM_005259075.3 XM_017027004.1 XM_017027005.1 XM_017027006.1 XM_017027007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5009703 c.52C>G p.L18V Chr.19: 43269380-43269380 on Build GRCh38
6085448 c.43T>A p.W15R Chr.19: 43269389-43269389 on Build GRCh38
3535245 c.33G>A p.Q11Q Chr.19: 43269399-43269399 on Build GRCh38
3535244 c.45G>A p.W15* Chr.19: 43269387-43269387 on Build GRCh38
5008506 c.64+1G>T p.? Chr.19: 43269367-43269367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373194039 Chr.19: 43269466-43269466 on Build GRCh38
rs774569354 Chr.19: 43269427-43269427 on Build GRCh38
rs780348567 Chr.19: 43269408-43269408 on Build GRCh38
rs147388768 Chr.19: 43269398-43269398 on Build GRCh38
rs140899532 Chr.19: 43269410-43269410 on Build GRCh38
rs8100909 Chr.19: 43269310-43269310 on Build GRCh38
rs200083247 Chr.19: 43269424-43269424 on Build GRCh38
rs775070639 Chr.19: 43269372-43269372 on Build GRCh38
rs746637584 Chr.19: 43269373-43269373 on Build GRCh38
rs142118461 Chr.19: 43269397-43269397 on Build GRCh38
rs767567313 Chr.19: 43269431-43269431 on Build GRCh38
rs777410416 Chr.19: 43269337-43269337 on Build GRCh38
rs530590607 Chr.19: 43269413-43269413 on Build GRCh38
rs8101191 Chr.19: 43269380-43269380 on Build GRCh38
rs8101210 Chr.19: 43269420-43269420 on Build GRCh38
rs772216436 Chr.19: 43269471-43269471 on Build GRCh38
rs536138360 Chr.19: 43269293-43269293 on Build GRCh38
rs749150042 Chr.19: 43269352-43269352 on Build GRCh38
rs757127741 Chr.19: 43269358-43269358 on Build GRCh38
rs1039003811 Chr.19: 43269383-43269383 on Build GRCh38
rs771600550 Chr.19: 43269369-43269369 on Build GRCh38
rs376166511 Chr.19: 43269470-43269470 on Build GRCh38
rs755126225 Chr.19: 43269390-43269390 on Build GRCh38
rs755924444 Chr.19: 43269336-43269336 on Build GRCh38
rs377691726 Chr.19: 43269453-43269453 on Build GRCh38
rs763532502 Chr.19: 43269425-43269425 on Build GRCh38
rs751739071 Chr.19: 43269385-43269385 on Build GRCh38
rs752994722 Chr.19: 43269433-43269433 on Build GRCh38
rs554287344 Chr.19: 43269468-43269468 on Build GRCh38
rs555721125 Chr.19: 43269298-43269298 on Build GRCh38
rs559295202 Chr.19: 43269384-43269384 on Build GRCh38
rs369519588 Chr.19: 43269443-43269443 on Build GRCh38
rs377427610 Chr.19: 43269401-43269401 on Build GRCh38
rs1013829426 Chr.19: 43269405-43269405 on Build GRCh38
rs750979375 Chr.19: 43269448-43269448 on Build GRCh38
rs764384157 Chr.19: 43269317-43269317 on Build GRCh38
rs757078118 Chr.19: 43269400-43269400 on Build GRCh38
rs963753201 Chr.19: 43269437-43269437 on Build GRCh38
rs371768798 Chr.19: 43269350-43269350 on Build GRCh38
rs1039441799 Chr.19: 43269339-43269339 on Build GRCh38
rs745826676 Chr.19: 43269403-43269403 on Build GRCh38
rs750254072 Chr.19: 43269444-43269444 on Build GRCh38
rs746586258 Chr.19: 43269414-43269414 on Build GRCh38
rs3179010 Chr.19: 43269474-43269474 on Build GRCh38
rs751863314 Chr.19: 43269335-43269335 on Build GRCh38
rs762299111 Chr.19: 43269379-43269379 on Build GRCh38
rs569490240 Chr.19: 43269455-43269455 on Build GRCh38
rs576916268 Chr.19: 43269366-43269366 on Build GRCh38
rs759673572 Chr.19: 43269430-43269430 on Build GRCh38
rs768183364 Chr.19: 43269419-43269419 on Build GRCh38
rs371905704 Chr.19: 43269327-43269327 on Build GRCh38
rs768393076 Chr.19: 43269375-43269375 on Build GRCh38
rs377122465 Chr.19: 43269442-43269442 on Build GRCh38
rs774335680 Chr.19: 43269339-43269339 on Build GRCh38
rs777100235 Chr.19: 43269319-43269319 on Build GRCh38
rs1058071 Chr.19: 43269447-43269447 on Build GRCh38
rs73037706 Chr.19: 43269422-43269422 on Build GRCh38
rs535803861 Chr.19: 43269445-43269445 on Build GRCh38
rs766645733 Chr.19: 43269334-43269334 on Build GRCh38
rs758569765 Chr.19: 43269328-43269328 on Build GRCh38
rs780672352 Chr.19: 43269450-43269450 on Build GRCh38
rs557060988 Chr.19: 43269362-43269362 on Build GRCh38
rs151046158 Chr.19: 43269326-43269326 on Build GRCh38
rs369027070 Chr.19: 43269456-43269456 on Build GRCh38
rs763090477 Chr.19: 43269382-43269382 on Build GRCh38

View Full Product Details