Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60928668-60928923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132A
Entrez Gene ID
54972
Gene Name
transmembrane protein 132A
Gene Aliases
GBP, HSPA5BP1
UniGene
Hs.118552
Gene Chromosome Location
Chr.11: 60924441-60937159 on Build GRCh38
Transcripts
NM_017870.3 NM_178031.2 XM_017017951.1 XM_017017952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229679 c.736C>T p.Q246* Chr.11: 60928830-60928830 on Build GRCh38
1127718 c.607G>T p.A203S Chr.11: 60928701-60928701 on Build GRCh38
3869677 c.697C>T p.P233S Chr.11: 60928791-60928791 on Build GRCh38
7133308 c.587C>T p.S196L Chr.11: 60928681-60928681 on Build GRCh38
2036678 c.702delG p.G236fs*21 Chr.11: 60928796-60928796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753895124 Chr.11: 60928688-60928688 on Build GRCh38
rs774876744 Chr.11: 60928756-60928756 on Build GRCh38
rs769032582 Chr.11: 60928864-60928865 on Build GRCh38
rs747327770 Chr.11: 60928833-60928833 on Build GRCh38
rs929716792 Chr.11: 60928717-60928717 on Build GRCh38
rs757144174 Chr.11: 60928691-60928691 on Build GRCh38
rs149956203 Chr.11: 60928761-60928761 on Build GRCh38
rs564681014 Chr.11: 60928703-60928703 on Build GRCh38
rs771375975 Chr.11: 60928744-60928744 on Build GRCh38
rs547896384 Chr.11: 60928777-60928777 on Build GRCh38
rs895161698 Chr.11: 60928788-60928788 on Build GRCh38
rs765416128 Chr.11: 60928862-60928862 on Build GRCh38
rs764212945 Chr.11: 60928687-60928687 on Build GRCh38
rs151190793 Chr.11: 60928698-60928698 on Build GRCh38
rs777345475 Chr.11: 60928891-60928891 on Build GRCh38
rs758396578 Chr.11: 60928772-60928772 on Build GRCh38
rs780846252 Chr.11: 60928798-60928798 on Build GRCh38
rs966912582 Chr.11: 60928835-60928835 on Build GRCh38
rs576673093 Chr.11: 60928870-60928870 on Build GRCh38
rs779963929 Chr.11: 60928701-60928701 on Build GRCh38
rs202064246 Chr.11: 60928757-60928757 on Build GRCh38
rs773160813 Chr.11: 60928864-60928864 on Build GRCh38
rs371465076 Chr.11: 60928704-60928704 on Build GRCh38
rs140281492 Chr.11: 60928890-60928890 on Build GRCh38
rs759978378 Chr.11: 60928758-60928758 on Build GRCh38
rs754688460 Chr.11: 60928797-60928797 on Build GRCh38
rs760897066 Chr.11: 60928760-60928760 on Build GRCh38
rs747630288 Chr.11: 60928718-60928718 on Build GRCh38
rs761919816 Chr.11: 60928856-60928856 on Build GRCh38
rs911126703 Chr.11: 60928694-60928694 on Build GRCh38
rs144893604 Chr.11: 60928769-60928769 on Build GRCh38
rs539239034 Chr.11: 60928887-60928887 on Build GRCh38
rs751419986 Chr.11: 60928871-60928871 on Build GRCh38
rs746726353 Chr.11: 60928702-60928702 on Build GRCh38
rs767181431 Chr.11: 60928882-60928882 on Build GRCh38
rs754776416 Chr.11: 60928874-60928874 on Build GRCh38
rs77946216 Chr.11: 60928812-60928812 on Build GRCh38
rs906122565 Chr.11: 60928811-60928811 on Build GRCh38
rs144698752 Chr.11: 60928853-60928853 on Build GRCh38
rs372811050 Chr.11: 60928896-60928896 on Build GRCh38
rs1032215731 Chr.11: 60928895-60928895 on Build GRCh38
rs751331854 Chr.11: 60928796-60928796 on Build GRCh38
rs140412965 Chr.11: 60928699-60928699 on Build GRCh38
rs536754543 Chr.11: 60928828-60928828 on Build GRCh38
rs140767642 Chr.11: 60928829-60928829 on Build GRCh38
rs747639573 Chr.11: 60928800-60928800 on Build GRCh38
rs762987035 Chr.11: 60928869-60928869 on Build GRCh38
rs761902925 Chr.11: 60928767-60928767 on Build GRCh38
rs370425430 Chr.11: 60928741-60928741 on Build GRCh38
rs374644513 Chr.11: 60928770-60928770 on Build GRCh38
rs369259825 Chr.11: 60928813-60928813 on Build GRCh38
rs749738974 Chr.11: 60928823-60928823 on Build GRCh38

View Full Product Details