Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 33778357-33778630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAS3
Entrez Gene ID
64067
Gene Name
neuronal PAS domain protein 3
Gene Aliases
MOP6, PASD6, bHLHe12
UniGene
Hs.657892
Gene Chromosome Location
Chr.14: 32934785-33804176 on Build GRCh38
Transcripts
NM_001164749.1 NM_001165893.1 NM_022123.2 NM_173159.2 XM_005267991.3 XM_005267992.3 XM_011537067.2 XM_011537069.2 XM_011537070.2 XM_011537071.2 XM_011537072.2 XM_017021582.1 XM_017021583.1 XM_017021584.1 XM_017021585.1 XM_017021586.1 XM_017021587.1 XM_017021588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
313325 c.1013G>T p.S338I Chr.14: 33778471-33778471 on Build GRCh38
6420826 c.1069A>T p.T357S Chr.14: 33778488-33778488 on Build GRCh38
6728494 c.991G>T p.G331W Chr.14: 33778506-33778506 on Build GRCh38
6420827 c.1030A>T p.T344S Chr.14: 33778488-33778488 on Build GRCh38
7289198 c.1062C>T p.Y354Y Chr.14: 33778520-33778520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374520771 Chr.14: 33778600-33778600 on Build GRCh38
rs778315395 Chr.14: 33778596-33778596 on Build GRCh38
rs549784028 Chr.14: 33778608-33778608 on Build GRCh38
rs747259085 Chr.14: 33778430-33778430 on Build GRCh38
rs927172585 Chr.14: 33778391-33778391 on Build GRCh38
rs762874557 Chr.14: 33778589-33778589 on Build GRCh38
rs141108484 Chr.14: 33778494-33778494 on Build GRCh38
rs112091405 Chr.14: 33778425-33778425 on Build GRCh38
rs3831102 Chr.14: 33778425-33778426 on Build GRCh38
rs397703018 Chr.14: 33778427-33778428 on Build GRCh38
rs781502728 Chr.14: 33778592-33778592 on Build GRCh38
rs750583541 Chr.14: 33778419-33778419 on Build GRCh38
rs780208181 Chr.14: 33778427-33778427 on Build GRCh38
rs753109616 Chr.14: 33778594-33778594 on Build GRCh38
rs758637169 Chr.14: 33778420-33778420 on Build GRCh38
rs562470607 Chr.14: 33778399-33778399 on Build GRCh38
rs749801737 Chr.14: 33778599-33778599 on Build GRCh38
rs373731568 Chr.14: 33778393-33778393 on Build GRCh38
rs115591737 Chr.14: 33778604-33778604 on Build GRCh38
rs759238078 Chr.14: 33778529-33778529 on Build GRCh38
rs184170938 Chr.14: 33778503-33778503 on Build GRCh38
rs762761601 Chr.14: 33778572-33778572 on Build GRCh38
rs756599367 Chr.14: 33778595-33778595 on Build GRCh38
rs772686768 Chr.14: 33778489-33778489 on Build GRCh38
rs778535422 Chr.14: 33778450-33778451 on Build GRCh38
rs760671254 Chr.14: 33778567-33778567 on Build GRCh38
rs770323066 Chr.14: 33778457-33778457 on Build GRCh38
rs148235325 Chr.14: 33778556-33778556 on Build GRCh38
rs764188832 Chr.14: 33778580-33778580 on Build GRCh38
rs138128927 Chr.14: 33778553-33778553 on Build GRCh38
rs755242466 Chr.14: 33778449-33778449 on Build GRCh38
rs968438331 Chr.14: 33778444-33778444 on Build GRCh38
rs202167736 Chr.14: 33778590-33778590 on Build GRCh38
rs781559920 Chr.14: 33778454-33778454 on Build GRCh38
rs773805152 Chr.14: 33778514-33778514 on Build GRCh38
rs538958798 Chr.14: 33778456-33778456 on Build GRCh38
rs549074013 Chr.14: 33778583-33778583 on Build GRCh38
rs147070037 Chr.14: 33778541-33778541 on Build GRCh38

View Full Product Details