Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5645212-5645485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAFB
Entrez Gene ID
6294
Gene Name
scaffold attachment factor B
Gene Aliases
HAP, HET, SAB-B1, SAF-B, SAF-B1, SAFB1
UniGene
Hs.728802
Gene Chromosome Location
Chr.19: 5623035-5668478 on Build GRCh38
Transcripts
NM_001201338.1 NM_001201339.1 NM_001201340.1 NM_001320571.1 NM_001320572.1 NM_002967.3 XM_017027114.1 XM_017027115.1 XM_017027116.1 XM_017027117.1 XM_017027118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501398 c.587C>T p.S196L Chr.19: 5645377-5645377 on Build GRCh38
1001737 c.558A>C p.K186N Chr.19: 5645348-5645348 on Build GRCh38
6771495 c.575T>G p.L192* Chr.19: 5645365-5645365 on Build GRCh38
5439898 c.600T>C p.T200T Chr.19: 5645390-5645390 on Build GRCh38
3423214 c.554A>G p.D185G Chr.19: 5645344-5645344 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755162887 Chr.19: 5645374-5645374 on Build GRCh38
rs775165023 Chr.19: 5645305-5645305 on Build GRCh38
rs1007943292 Chr.19: 5645238-5645238 on Build GRCh38
rs200977093 Chr.19: 5645366-5645366 on Build GRCh38
rs200372829 Chr.19: 5645391-5645391 on Build GRCh38
rs1000721035 Chr.19: 5645308-5645308 on Build GRCh38
rs184786649 Chr.19: 5645437-5645437 on Build GRCh38
rs2436502 Chr.19: 5645433-5645433 on Build GRCh38
rs751748598 Chr.19: 5645371-5645371 on Build GRCh38
rs761480856 Chr.19: 5645332-5645332 on Build GRCh38
rs752710076 Chr.19: 5645389-5645389 on Build GRCh38
rs771890559 Chr.19: 5645302-5645302 on Build GRCh38
rs373455700 Chr.19: 5645402-5645402 on Build GRCh38
rs758137846 Chr.19: 5645394-5645394 on Build GRCh38
rs768187540 Chr.19: 5645415-5645415 on Build GRCh38
rs767849744 Chr.19: 5645378-5645378 on Build GRCh38
rs763370611 Chr.19: 5645365-5645365 on Build GRCh38
rs76200723 Chr.19: 5645392-5645392 on Build GRCh38
rs879906456 Chr.19: 5645254-5645254 on Build GRCh38
rs778883634 Chr.19: 5645290-5645290 on Build GRCh38
rs780954790 Chr.19: 5645419-5645419 on Build GRCh38
rs373422884 Chr.19: 5645447-5645447 on Build GRCh38
rs745644707 Chr.19: 5645291-5645291 on Build GRCh38
rs776261119 Chr.19: 5645295-5645297 on Build GRCh38
rs993738184 Chr.19: 5645316-5645316 on Build GRCh38
rs772279680 Chr.19: 5645440-5645440 on Build GRCh38
rs543811055 Chr.19: 5645411-5645411 on Build GRCh38
rs765825547 Chr.19: 5645341-5645341 on Build GRCh38
rs776008074 Chr.19: 5645330-5645330 on Build GRCh38
rs1043151881 Chr.19: 5645251-5645251 on Build GRCh38
rs773904938 Chr.19: 5645342-5645342 on Build GRCh38
rs192764954 Chr.19: 5645432-5645432 on Build GRCh38
rs555880346 Chr.19: 5645278-5645278 on Build GRCh38
rs970885098 Chr.19: 5645375-5645375 on Build GRCh38
rs768152110 Chr.19: 5645318-5645318 on Build GRCh38
rs150067581 Chr.19: 5645310-5645310 on Build GRCh38

View Full Product Details