Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3198091-3198358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1A2
Entrez Gene ID
26189
Gene Name
olfactory receptor family 1 subfamily A member 2
Gene Aliases
OR17-6
UniGene
Hs.532660
Gene Chromosome Location
Chr.17: 3197517-3198448 on Build GRCh38
Transcripts
NM_012352.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5386040 c.618C>T p.F206F Chr.17: 3198136-3198136 on Build GRCh38
1679379 c.650A>G p.Y217C Chr.17: 3198168-3198168 on Build GRCh38
5425324 c.766G>T p.G256C Chr.17: 3198284-3198284 on Build GRCh38
1709568 c.752A>G p.Y251C Chr.17: 3198270-3198270 on Build GRCh38
5625171 c.648C>G p.S216S Chr.17: 3198166-3198166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781401043 Chr.17: 3198257-3198258 on Build GRCh38
rs367950308 Chr.17: 3198262-3198262 on Build GRCh38
rs767481087 Chr.17: 3198271-3198271 on Build GRCh38
rs747460000 Chr.17: 3198306-3198306 on Build GRCh38
rs775775400 Chr.17: 3198230-3198230 on Build GRCh38
rs75713349 Chr.17: 3198279-3198279 on Build GRCh38
rs374656880 Chr.17: 3198257-3198257 on Build GRCh38
rs755910705 Chr.17: 3198280-3198280 on Build GRCh38
rs780886513 Chr.17: 3198178-3198178 on Build GRCh38
rs199681281 Chr.17: 3198226-3198226 on Build GRCh38
rs762731445 Chr.17: 3198326-3198326 on Build GRCh38
rs367689045 Chr.17: 3198232-3198232 on Build GRCh38
rs778900556 Chr.17: 3198287-3198287 on Build GRCh38
rs774442114 Chr.17: 3198213-3198213 on Build GRCh38
rs200511075 Chr.17: 3198187-3198187 on Build GRCh38
rs754901352 Chr.17: 3198169-3198169 on Build GRCh38
rs757432372 Chr.17: 3198286-3198286 on Build GRCh38
rs2469791 Chr.17: 3198296-3198296 on Build GRCh38
rs746394716 Chr.17: 3198215-3198215 on Build GRCh38
rs770591576 Chr.17: 3198336-3198336 on Build GRCh38
rs769162911 Chr.17: 3198310-3198310 on Build GRCh38
rs747949056 Chr.17: 3198183-3198183 on Build GRCh38
rs189449756 Chr.17: 3198204-3198204 on Build GRCh38
rs111855057 Chr.17: 3198297-3198297 on Build GRCh38
rs2241091 Chr.17: 3198284-3198284 on Build GRCh38
rs765931476 Chr.17: 3198151-3198151 on Build GRCh38
rs745580713 Chr.17: 3198191-3198197 on Build GRCh38
rs965044370 Chr.17: 3198138-3198138 on Build GRCh38
rs924678778 Chr.17: 3198162-3198162 on Build GRCh38
rs751472380 Chr.17: 3198152-3198152 on Build GRCh38
rs754083813 Chr.17: 3198285-3198285 on Build GRCh38
rs150553361 Chr.17: 3198127-3198127 on Build GRCh38
rs764970882 Chr.17: 3198140-3198140 on Build GRCh38
rs978338995 Chr.17: 3198154-3198154 on Build GRCh38
rs771545111 Chr.17: 3198129-3198129 on Build GRCh38
rs771380407 Chr.17: 3198161-3198187 on Build GRCh38
rs764485219 Chr.17: 3198247-3198247 on Build GRCh38
rs373842311 Chr.17: 3198131-3198131 on Build GRCh38
rs1006440504 Chr.17: 3198130-3198130 on Build GRCh38
rs771013870 Chr.17: 3198211-3198211 on Build GRCh38
rs757987502 Chr.17: 3198148-3198148 on Build GRCh38
rs747206669 Chr.17: 3198126-3198126 on Build GRCh38
rs184251188 Chr.17: 3198186-3198186 on Build GRCh38
rs56058341 Chr.17: 3198248-3198248 on Build GRCh38
rs777093479 Chr.17: 3198321-3198321 on Build GRCh38
rs780615147 Chr.17: 3198305-3198305 on Build GRCh38
rs750021378 Chr.17: 3198141-3198141 on Build GRCh38

View Full Product Details