Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219281862-219282129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4001514 c.230-10G>A p.? Chr.2: 219281929-219281929 on Build GRCh38
6900869 c.296C>T p.P99L Chr.2: 219282005-219282005 on Build GRCh38
3577948 c.309C>T p.F103F Chr.2: 219282018-219282018 on Build GRCh38
3682543 c.230-10G>C p.? Chr.2: 219281929-219281929 on Build GRCh38
6157009 c.233C>T p.T78I Chr.2: 219281942-219281942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376497742 Chr.2: 219282012-219282012 on Build GRCh38
rs749512177 Chr.2: 219281969-219281969 on Build GRCh38
rs778179196 Chr.2: 219281892-219281892 on Build GRCh38
rs749000825 Chr.2: 219282104-219282104 on Build GRCh38
rs763591754 Chr.2: 219282032-219282032 on Build GRCh38
rs371501438 Chr.2: 219281914-219281914 on Build GRCh38
rs750840887 Chr.2: 219281964-219281964 on Build GRCh38
rs756614404 Chr.2: 219282062-219282062 on Build GRCh38
rs755259886 Chr.2: 219282081-219282081 on Build GRCh38
rs376990198 Chr.2: 219282110-219282110 on Build GRCh38
rs770705479 Chr.2: 219281921-219281921 on Build GRCh38
rs778951823 Chr.2: 219282084-219282084 on Build GRCh38
rs1037119110 Chr.2: 219281899-219281899 on Build GRCh38
rs369661561 Chr.2: 219281937-219281937 on Build GRCh38
rs576058376 Chr.2: 219281958-219281958 on Build GRCh38
rs546107193 Chr.2: 219281915-219281915 on Build GRCh38
rs748304935 Chr.2: 219282093-219282093 on Build GRCh38
rs892194753 Chr.2: 219281885-219281885 on Build GRCh38
rs762434804 Chr.2: 219281949-219281949 on Build GRCh38
rs779054411 Chr.2: 219281997-219281997 on Build GRCh38
rs1049568759 Chr.2: 219281882-219281883 on Build GRCh38
rs145429721 Chr.2: 219282020-219282020 on Build GRCh38
rs989258467 Chr.2: 219282069-219282069 on Build GRCh38
rs200449308 Chr.2: 219282097-219282097 on Build GRCh38
rs3832110 Chr.2: 219281887-219281888 on Build GRCh38
rs561000982 Chr.2: 219282095-219282095 on Build GRCh38
rs545130842 Chr.2: 219281999-219281999 on Build GRCh38
rs370349276 Chr.2: 219282065-219282065 on Build GRCh38
rs754082602 Chr.2: 219282067-219282067 on Build GRCh38
rs769569269 Chr.2: 219282010-219282010 on Build GRCh38
rs757624144 Chr.2: 219281907-219281907 on Build GRCh38
rs756526573 Chr.2: 219281967-219281967 on Build GRCh38
rs775361970 Chr.2: 219282100-219282100 on Build GRCh38
rs146284616 Chr.2: 219281918-219281918 on Build GRCh38
rs780465205 Chr.2: 219281968-219281968 on Build GRCh38
rs3731897 Chr.2: 219281929-219281929 on Build GRCh38
rs768700590 Chr.2: 219281987-219281987 on Build GRCh38
rs775372866 Chr.2: 219281945-219281945 on Build GRCh38
rs375885717 Chr.2: 219281916-219281916 on Build GRCh38
rs776440511 Chr.2: 219281924-219281924 on Build GRCh38
rs1050045878 Chr.2: 219282051-219282051 on Build GRCh38
rs769665024 Chr.2: 219282099-219282099 on Build GRCh38
rs750930950 Chr.2: 219282056-219282056 on Build GRCh38
rs764916088 Chr.2: 219281934-219281934 on Build GRCh38
rs1029212324 Chr.2: 219281920-219281920 on Build GRCh38
rs397783034 Chr.2: 219281888-219281889 on Build GRCh38
rs761180152 Chr.2: 219282042-219282042 on Build GRCh38
rs576205050 Chr.2: 219281954-219281954 on Build GRCh38
rs746171813 Chr.2: 219281913-219281913 on Build GRCh38
rs202090561 Chr.2: 219281887-219281887 on Build GRCh38
rs139406455 Chr.2: 219282007-219282007 on Build GRCh38
rs747550577 Chr.2: 219281894-219281894 on Build GRCh38
rs751116201 Chr.2: 219282040-219282040 on Build GRCh38
rs541289584 Chr.2: 219282054-219282054 on Build GRCh38
rs373432472 Chr.2: 219282006-219282006 on Build GRCh38
rs879253868 Chr.2: 219282018-219282018 on Build GRCh38
rs797045039 Chr.2: 219282052-219282052 on Build GRCh38
rs763649673 Chr.2: 219281953-219281953 on Build GRCh38
rs367625569 Chr.2: 219281909-219281909 on Build GRCh38

View Full Product Details