Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47143785-47144018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4A22
Entrez Gene ID
284541
Gene Name
cytochrome P450 family 4 subfamily A member 22
Gene Aliases
-
UniGene
Hs.726474
Gene Chromosome Location
Chr.1: 47137425-47149738 on Build GRCh38
Transcripts
NM_001010969.3 NM_001308102.1 XM_005270767.3 XM_005270770.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760290 c.737C>G p.T246S Chr.1: 47143863-47143863 on Build GRCh38
5053554 c.745G>T p.G249C Chr.1: 47143871-47143871 on Build GRCh38
7084337 c.696G>C p.M232I Chr.1: 47143822-47143822 on Build GRCh38
3746998 c.691T>C p.C231R Chr.1: 47143817-47143817 on Build GRCh38
6127042 c.784C>A p.H262N Chr.1: 47143910-47143910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143425502 Chr.1: 47143844-47143844 on Build GRCh38
rs750494406 Chr.1: 47143966-47143966 on Build GRCh38
rs550279903 Chr.1: 47143859-47143859 on Build GRCh38
rs770005682 Chr.1: 47143850-47143850 on Build GRCh38
rs772472507 Chr.1: 47143922-47143922 on Build GRCh38
rs765043322 Chr.1: 47143874-47143874 on Build GRCh38
rs1040351534 Chr.1: 47143969-47143969 on Build GRCh38
rs749710175 Chr.1: 47143853-47143853 on Build GRCh38
rs762123692 Chr.1: 47143953-47143953 on Build GRCh38
rs781263992 Chr.1: 47143890-47143890 on Build GRCh38
rs750514702 Chr.1: 47143891-47143891 on Build GRCh38
rs2405590 Chr.1: 47143926-47143926 on Build GRCh38
rs757407453 Chr.1: 47143838-47143838 on Build GRCh38
rs752556492 Chr.1: 47143829-47143829 on Build GRCh38
rs199906672 Chr.1: 47143851-47143851 on Build GRCh38
rs763912452 Chr.1: 47143949-47143949 on Build GRCh38
rs777837250 Chr.1: 47143836-47143836 on Build GRCh38
rs191687330 Chr.1: 47143820-47143820 on Build GRCh38
rs765912026 Chr.1: 47143818-47143818 on Build GRCh38
rs760078696 Chr.1: 47143863-47143863 on Build GRCh38
rs780321461 Chr.1: 47143900-47143900 on Build GRCh38
rs376758503 Chr.1: 47143889-47143889 on Build GRCh38
rs982039925 Chr.1: 47143905-47143905 on Build GRCh38
rs746052339 Chr.1: 47143845-47143845 on Build GRCh38
rs769112812 Chr.1: 47143909-47143909 on Build GRCh38
rs758308317 Chr.1: 47143878-47143878 on Build GRCh38
rs772583047 Chr.1: 47143808-47143808 on Build GRCh38
rs764597121 Chr.1: 47143807-47143807 on Build GRCh38
rs775336205 Chr.1: 47143944-47143944 on Build GRCh38
rs777023100 Chr.1: 47143949-47143949 on Build GRCh38
rs769147059 Chr.1: 47143857-47143857 on Build GRCh38
rs759209238 Chr.1: 47143873-47143873 on Build GRCh38
rs536116490 Chr.1: 47143887-47143887 on Build GRCh38
rs35156123 Chr.1: 47143815-47143815 on Build GRCh38
rs763383232 Chr.1: 47143806-47143806 on Build GRCh38
rs754698284 Chr.1: 47143825-47143825 on Build GRCh38
rs756315173 Chr.1: 47143892-47143892 on Build GRCh38
rs113636046 Chr.1: 47143935-47143935 on Build GRCh38
rs558608539 Chr.1: 47143975-47143975 on Build GRCh38
rs10789501 Chr.1: 47143817-47143817 on Build GRCh38
rs183880611 Chr.1: 47143832-47143832 on Build GRCh38
rs35609683 Chr.1: 47143960-47143960 on Build GRCh38
rs147982626 Chr.1: 47143870-47143870 on Build GRCh38
rs767409990 Chr.1: 47143962-47143962 on Build GRCh38
rs921552200 Chr.1: 47143986-47143986 on Build GRCh38
rs747028616 Chr.1: 47143837-47143837 on Build GRCh38
rs765059748 Chr.1: 47143827-47143827 on Build GRCh38
rs911909 Chr.1: 47143996-47143996 on Build GRCh38
rs554345540 Chr.1: 47143901-47143901 on Build GRCh38
rs138579860 Chr.1: 47143880-47143880 on Build GRCh38
rs569148380 Chr.1: 47143875-47143875 on Build GRCh38
rs147311489 Chr.1: 47143907-47143907 on Build GRCh38
rs374540641 Chr.1: 47143888-47143888 on Build GRCh38
rs779166657 Chr.1: 47143914-47143914 on Build GRCh38
rs751319143 Chr.1: 47143951-47143951 on Build GRCh38
rs769540925 Chr.1: 47143936-47143936 on Build GRCh38
rs765861938 Chr.1: 47143864-47143864 on Build GRCh38
rs112811686 Chr.1: 47143917-47143917 on Build GRCh38

View Full Product Details