Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70979246-70979471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD53
Entrez Gene ID
79998
Gene Name
ankyrin repeat domain 53
Gene Aliases
-
UniGene
Hs.512744
Gene Chromosome Location
Chr.2: 70977183-70985499 on Build GRCh38
Transcripts
NM_001115116.1 NM_024933.3 XM_005264569.4 XM_011533114.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669014 c.323G>T p.S108I Chr.2: 70979249-70979249 on Build GRCh38
6856211 c.345G>A p.A115A Chr.2: 70979271-70979271 on Build GRCh38
5627509 c.414_415insA p.F141fs*34 Chr.2: 70979340-70979341 on Build GRCh38
7136877 c.384C>A p.N128K Chr.2: 70979310-70979310 on Build GRCh38
5973847 c.359T>C p.V120A Chr.2: 70979285-70979285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782270006 Chr.2: 70979292-70979292 on Build GRCh38
rs782225477 Chr.2: 70979285-70979285 on Build GRCh38
rs782754325 Chr.2: 70979387-70979387 on Build GRCh38
rs782645878 Chr.2: 70979287-70979287 on Build GRCh38
rs377019890 Chr.2: 70979359-70979359 on Build GRCh38
rs1002104412 Chr.2: 70979342-70979342 on Build GRCh38
rs781811070 Chr.2: 70979379-70979379 on Build GRCh38
rs782590629 Chr.2: 70979283-70979283 on Build GRCh38
rs544496853 Chr.2: 70979420-70979420 on Build GRCh38
rs781967323 Chr.2: 70979337-70979337 on Build GRCh38
rs371048286 Chr.2: 70979392-70979392 on Build GRCh38
rs904646865 Chr.2: 70979312-70979312 on Build GRCh38
rs375062592 Chr.2: 70979338-70979338 on Build GRCh38
rs782412700 Chr.2: 70979302-70979302 on Build GRCh38
rs781906105 Chr.2: 70979275-70979275 on Build GRCh38
rs782087531 Chr.2: 70979386-70979386 on Build GRCh38
rs567346526 Chr.2: 70979442-70979442 on Build GRCh38
rs781916885 Chr.2: 70979305-70979305 on Build GRCh38
rs147026593 Chr.2: 70979271-70979271 on Build GRCh38
rs1024568842 Chr.2: 70979399-70979399 on Build GRCh38
rs782165379 Chr.2: 70979363-70979363 on Build GRCh38
rs549048154 Chr.2: 70979441-70979441 on Build GRCh38
rs782455845 Chr.2: 70979276-70979276 on Build GRCh38
rs782498592 Chr.2: 70979286-70979286 on Build GRCh38
rs575192264 Chr.2: 70979306-70979306 on Build GRCh38
rs1014478696 Chr.2: 70979398-70979398 on Build GRCh38
rs782783901 Chr.2: 70979344-70979344 on Build GRCh38
rs782696296 Chr.2: 70979378-70979378 on Build GRCh38
rs781863930 Chr.2: 70979390-70979390 on Build GRCh38
rs782190864 Chr.2: 70979333-70979333 on Build GRCh38
rs961008946 Chr.2: 70979424-70979424 on Build GRCh38
rs782342287 Chr.2: 70979335-70979335 on Build GRCh38
rs782678207 Chr.2: 70979394-70979394 on Build GRCh38
rs1034532675 Chr.2: 70979343-70979343 on Build GRCh38

View Full Product Details