Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207131371-207131618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPA
Entrez Gene ID
722
Gene Name
complement component 4 binding protein alpha
Gene Aliases
C4BP, PRP
UniGene
Hs.1012
Gene Chromosome Location
Chr.1: 207104232-207144972 on Build GRCh38
Transcripts
NM_000715.3 XM_005273251.1 XM_005273252.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180276 c.920A>T p.H307L Chr.1: 207131576-207131576 on Build GRCh38
678890 c.940C>T p.P314S Chr.1: 207131596-207131596 on Build GRCh38
4573030 c.942T>C p.P314P Chr.1: 207131598-207131598 on Build GRCh38
7277901 c.948G>A p.P316P Chr.1: 207131604-207131604 on Build GRCh38
3482597 c.922G>A p.A308T Chr.1: 207131578-207131578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779229160 Chr.1: 207131506-207131506 on Build GRCh38
rs141556894 Chr.1: 207131519-207131521 on Build GRCh38
rs762343207 Chr.1: 207131572-207131572 on Build GRCh38
rs1003263079 Chr.1: 207131441-207131441 on Build GRCh38
rs199696184 Chr.1: 207131580-207131580 on Build GRCh38
rs781197219 Chr.1: 207131497-207131497 on Build GRCh38
rs961544894 Chr.1: 207131487-207131487 on Build GRCh38
rs956582853 Chr.1: 207131405-207131405 on Build GRCh38
rs775226640 Chr.1: 207131571-207131571 on Build GRCh38
rs775971186 Chr.1: 207131521-207131521 on Build GRCh38
rs4844573 Chr.1: 207131555-207131555 on Build GRCh38
rs1014011771 Chr.1: 207131422-207131422 on Build GRCh38
rs142864153 Chr.1: 207131553-207131553 on Build GRCh38
rs749505213 Chr.1: 207131498-207131498 on Build GRCh38
rs200240900 Chr.1: 207131510-207131510 on Build GRCh38
rs751538346 Chr.1: 207131582-207131582 on Build GRCh38
rs765977791 Chr.1: 207131518-207131518 on Build GRCh38
rs906270714 Chr.1: 207131429-207131429 on Build GRCh38
rs539147120 Chr.1: 207131509-207131509 on Build GRCh38
rs963579121 Chr.1: 207131523-207131523 on Build GRCh38
rs764753591 Chr.1: 207131558-207131558 on Build GRCh38
rs534602199 Chr.1: 207131556-207131556 on Build GRCh38
rs747585918 Chr.1: 207131512-207131512 on Build GRCh38
rs558290223 Chr.1: 207131503-207131504 on Build GRCh38
rs755095539 Chr.1: 207131500-207131500 on Build GRCh38
rs570130802 Chr.1: 207131451-207131451 on Build GRCh38

View Full Product Details