Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203223090-203223351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
UniGene
Hs.201688
Gene Chromosome Location
Chr.1: 203216079-203229732 on Build GRCh38
Transcripts
NM_001256125.1 NM_001270509.1 NM_003465.2 NR_045784.1 NR_045785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3481852 c.521G>A p.G174E Chr.1: 203223219-203223219 on Build GRCh38
7135830 c.573C>A p.I191I Chr.1: 203223140-203223140 on Build GRCh38
365472 c.601G>A p.A201T Chr.1: 203223139-203223139 on Build GRCh38
6032461 c.589G>A p.V197M Chr.1: 203223151-203223151 on Build GRCh38
678071 c.554C>A p.P185Q Chr.1: 203223186-203223186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188646454 Chr.1: 203223278-203223278 on Build GRCh38
rs762479668 Chr.1: 203223282-203223282 on Build GRCh38
rs749270846 Chr.1: 203223146-203223146 on Build GRCh38
rs757224498 Chr.1: 203223140-203223140 on Build GRCh38
rs773755505 Chr.1: 203223186-203223186 on Build GRCh38
rs777287951 Chr.1: 203223197-203223197 on Build GRCh38
rs771422197 Chr.1: 203223257-203223257 on Build GRCh38
rs150370762 Chr.1: 203223139-203223139 on Build GRCh38
rs760147105 Chr.1: 203223261-203223261 on Build GRCh38
rs764864572 Chr.1: 203223207-203223207 on Build GRCh38
rs751220019 Chr.1: 203223294-203223294 on Build GRCh38
rs943587511 Chr.1: 203223254-203223254 on Build GRCh38
rs369795637 Chr.1: 203223247-203223247 on Build GRCh38
rs574974603 Chr.1: 203223154-203223154 on Build GRCh38
rs114168492 Chr.1: 203223155-203223155 on Build GRCh38
rs768738986 Chr.1: 203223120-203223121 on Build GRCh38
rs200777680 Chr.1: 203223211-203223211 on Build GRCh38
rs754999693 Chr.1: 203223226-203223226 on Build GRCh38
rs140770731 Chr.1: 203223162-203223162 on Build GRCh38
rs529879573 Chr.1: 203223314-203223314 on Build GRCh38
rs776595649 Chr.1: 203223201-203223203 on Build GRCh38
rs535047884 Chr.1: 203223142-203223142 on Build GRCh38
rs151004649 Chr.1: 203223232-203223232 on Build GRCh38
rs758258937 Chr.1: 203223184-203223184 on Build GRCh38
rs367730457 Chr.1: 203223183-203223183 on Build GRCh38
rs147465832 Chr.1: 203223239-203223239 on Build GRCh38
rs776996836 Chr.1: 203223260-203223260 on Build GRCh38
rs200929749 Chr.1: 203223293-203223293 on Build GRCh38
rs12562058 Chr.1: 203223227-203223227 on Build GRCh38
rs3216011 Chr.1: 203223321-203223321 on Build GRCh38
rs368307246 Chr.1: 203223125-203223125 on Build GRCh38
rs372266737 Chr.1: 203223171-203223171 on Build GRCh38
rs577584015 Chr.1: 203223215-203223215 on Build GRCh38
rs377276949 Chr.1: 203223192-203223192 on Build GRCh38
rs376557890 Chr.1: 203223295-203223295 on Build GRCh38
rs181385947 Chr.1: 203223296-203223296 on Build GRCh38
rs928517162 Chr.1: 203223307-203223307 on Build GRCh38
rs397827197 Chr.1: 203223324-203223324 on Build GRCh38
rs555993709 Chr.1: 203223170-203223170 on Build GRCh38
rs777602913 Chr.1: 203223133-203223133 on Build GRCh38
rs745884865 Chr.1: 203223224-203223224 on Build GRCh38
rs752277107 Chr.1: 203223209-203223209 on Build GRCh38
rs752395878 Chr.1: 203223124-203223124 on Build GRCh38
rs201640938 Chr.1: 203223276-203223276 on Build GRCh38
rs781043062 Chr.1: 203223220-203223220 on Build GRCh38
rs747317143 Chr.1: 203223253-203223253 on Build GRCh38
rs764631127 Chr.1: 203223274-203223274 on Build GRCh38
rs371464440 Chr.1: 203223191-203223191 on Build GRCh38
rs758118986 Chr.1: 203223210-203223210 on Build GRCh38
rs756122031 Chr.1: 203223308-203223308 on Build GRCh38
rs762034144 Chr.1: 203223248-203223249 on Build GRCh38

View Full Product Details