Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5515854-5516125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBQLNL
Entrez Gene ID
143630
Gene Name
ubiquilin-like
Gene Aliases
-
UniGene
Hs.10688
Gene Chromosome Location
Chr.11: 5514393-5516726 on Build GRCh38
Transcripts
NM_145053.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306170 c.475G>C p.V159L Chr.11: 5515967-5515967 on Build GRCh38
7229058 c.427C>A p.Q143K Chr.11: 5516015-5516015 on Build GRCh38
4423423 c.415A>T p.T139S Chr.11: 5516027-5516027 on Build GRCh38
3869379 c.570C>T p.F190F Chr.11: 5515872-5515872 on Build GRCh38
1703469 c.439G>A p.E147K Chr.11: 5516003-5516003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144353879 Chr.11: 5516079-5516079 on Build GRCh38
rs376579289 Chr.11: 5516071-5516071 on Build GRCh38
rs745803586 Chr.11: 5516102-5516102 on Build GRCh38
rs2047456 Chr.11: 5515931-5515931 on Build GRCh38
rs3802978 Chr.11: 5516015-5516015 on Build GRCh38
rs752342835 Chr.11: 5516087-5516087 on Build GRCh38
rs201147545 Chr.11: 5515895-5515895 on Build GRCh38
rs368343827 Chr.11: 5516088-5516088 on Build GRCh38
rs754536037 Chr.11: 5515978-5515978 on Build GRCh38
rs753079827 Chr.11: 5515883-5515883 on Build GRCh38
rs137908718 Chr.11: 5515953-5515953 on Build GRCh38
rs148278465 Chr.11: 5515917-5515917 on Build GRCh38
rs753024488 Chr.11: 5515943-5515943 on Build GRCh38
rs751042264 Chr.11: 5515973-5515973 on Build GRCh38
rs765714697 Chr.11: 5516058-5516058 on Build GRCh38
rs769572435 Chr.11: 5516000-5516000 on Build GRCh38
rs780812097 Chr.11: 5515984-5515984 on Build GRCh38
rs771334765 Chr.11: 5515989-5515989 on Build GRCh38
rs755269814 Chr.11: 5515878-5515878 on Build GRCh38
rs191943572 Chr.11: 5516005-5516005 on Build GRCh38
rs757859792 Chr.11: 5515889-5515889 on Build GRCh38
rs146180638 Chr.11: 5515946-5515946 on Build GRCh38
rs770990230 Chr.11: 5515925-5515925 on Build GRCh38
rs199758059 Chr.11: 5515918-5515918 on Build GRCh38
rs770960057 Chr.11: 5516025-5516025 on Build GRCh38
rs764471232 Chr.11: 5516046-5516046 on Build GRCh38
rs757728031 Chr.11: 5515971-5515971 on Build GRCh38
rs878973356 Chr.11: 5516081-5516081 on Build GRCh38
rs564437441 Chr.11: 5516101-5516101 on Build GRCh38
rs1024281401 Chr.11: 5516052-5516052 on Build GRCh38
rs549099023 Chr.11: 5516026-5516026 on Build GRCh38
rs777518193 Chr.11: 5516089-5516089 on Build GRCh38
rs749013269 Chr.11: 5516099-5516099 on Build GRCh38
rs904354744 Chr.11: 5515922-5515922 on Build GRCh38
rs767953254 Chr.11: 5515881-5515881 on Build GRCh38
rs778328735 Chr.11: 5515888-5515888 on Build GRCh38
rs754186917 Chr.11: 5516047-5516047 on Build GRCh38
rs150539325 Chr.11: 5516036-5516036 on Build GRCh38
rs557573869 Chr.11: 5515894-5515894 on Build GRCh38
rs764371851 Chr.11: 5515949-5515949 on Build GRCh38
rs143659852 Chr.11: 5516072-5516072 on Build GRCh38
rs776310609 Chr.11: 5515906-5515906 on Build GRCh38
rs756482498 Chr.11: 5515886-5515886 on Build GRCh38
rs922286477 Chr.11: 5515912-5515912 on Build GRCh38
rs759673116 Chr.11: 5516031-5516031 on Build GRCh38
rs759729325 Chr.11: 5515930-5515930 on Build GRCh38
rs768332932 Chr.11: 5515899-5515899 on Build GRCh38
rs774600138 Chr.11: 5515928-5515928 on Build GRCh38
rs779500730 Chr.11: 5515972-5515972 on Build GRCh38
rs747955342 Chr.11: 5515914-5515914 on Build GRCh38
rs775789850 Chr.11: 5516039-5516039 on Build GRCh38
rs139342517 Chr.11: 5516045-5516045 on Build GRCh38
rs774479230 Chr.11: 5516029-5516029 on Build GRCh38
rs747825146 Chr.11: 5515997-5515997 on Build GRCh38
rs553557959 Chr.11: 5515944-5515944 on Build GRCh38
rs368898863 Chr.11: 5516020-5516020 on Build GRCh38

View Full Product Details