Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172770437-172770706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP1
Entrez Gene ID
1843
Gene Name
dual specificity phosphatase 1
Gene Aliases
CL100, HVH1, MKP-1, MKP1, PTPN10
UniGene
Hs.171695
Gene Chromosome Location
Chr.5: 172768090-172771200 on Build GRCh38
Transcripts
NM_004417.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964222 c.340C>T p.R114C Chr.5: 172770613-172770613 on Build GRCh38
5427058 c.367+8C>G p.? Chr.5: 172770578-172770578 on Build GRCh38
6650284 c.367+4C>T p.? Chr.5: 172770582-172770582 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006353250 Chr.5: 172770534-172770534 on Build GRCh38
rs569788061 Chr.5: 172770566-172770566 on Build GRCh38
rs13177271 Chr.5: 172770462-172770462 on Build GRCh38
rs1056835437 Chr.5: 172770643-172770643 on Build GRCh38
rs752430292 Chr.5: 172770556-172770556 on Build GRCh38
rs961127280 Chr.5: 172770547-172770547 on Build GRCh38
rs561090164 Chr.5: 172770482-172770482 on Build GRCh38
rs769037461 Chr.5: 172770584-172770584 on Build GRCh38
rs547362788 Chr.5: 172770465-172770465 on Build GRCh38
rs922298779 Chr.5: 172770565-172770565 on Build GRCh38
rs375185075 Chr.5: 172770574-172770574 on Build GRCh38
rs972779265 Chr.5: 172770552-172770552 on Build GRCh38
rs1024186981 Chr.5: 172770505-172770505 on Build GRCh38
rs773968338 Chr.5: 172770608-172770608 on Build GRCh38
rs34332995 Chr.5: 172770532-172770532 on Build GRCh38
rs755096464 Chr.5: 172770645-172770645 on Build GRCh38
rs745364103 Chr.5: 172770536-172770536 on Build GRCh38
rs781232951 Chr.5: 172770548-172770548 on Build GRCh38
rs778935204 Chr.5: 172770573-172770573 on Build GRCh38
rs1012703295 Chr.5: 172770681-172770681 on Build GRCh38
rs774531764 Chr.5: 172770545-172770545 on Build GRCh38
rs781488618 Chr.5: 172770650-172770650 on Build GRCh38
rs780317510 Chr.5: 172770629-172770629 on Build GRCh38
rs766186850 Chr.5: 172770605-172770605 on Build GRCh38
rs753682011 Chr.5: 172770621-172770621 on Build GRCh38
rs767344846 Chr.5: 172770615-172770615 on Build GRCh38
rs1032074063 Chr.5: 172770649-172770649 on Build GRCh38
rs772964123 Chr.5: 172770598-172770598 on Build GRCh38
rs896490406 Chr.5: 172770654-172770654 on Build GRCh38
rs762721199 Chr.5: 172770603-172770603 on Build GRCh38
rs754944959 Chr.5: 172770543-172770543 on Build GRCh38
rs549721126 Chr.5: 172770558-172770558 on Build GRCh38
rs778771698 Chr.5: 172770511-172770511 on Build GRCh38
rs759155703 Chr.5: 172770611-172770611 on Build GRCh38
rs966914280 Chr.5: 172770638-172770638 on Build GRCh38
rs763271149 Chr.5: 172770513-172770513 on Build GRCh38
rs770203276 Chr.5: 172770593-172770593 on Build GRCh38
rs747129421 Chr.5: 172770579-172770579 on Build GRCh38
rs946518552 Chr.5: 172770590-172770590 on Build GRCh38
rs748723790 Chr.5: 172770655-172770655 on Build GRCh38
rs552467759 Chr.5: 172770622-172770622 on Build GRCh38
rs1020876628 Chr.5: 172770634-172770634 on Build GRCh38
rs1012510072 Chr.5: 172770464-172770464 on Build GRCh38
rs748400009 Chr.5: 172770589-172770589 on Build GRCh38
rs777249380 Chr.5: 172770588-172770588 on Build GRCh38
rs752683966 Chr.5: 172770619-172770619 on Build GRCh38
rs755823321 Chr.5: 172770620-172770620 on Build GRCh38
rs923998270 Chr.5: 172770628-172770628 on Build GRCh38
rs770009927 Chr.5: 172770682-172770682 on Build GRCh38
rs747076239 Chr.5: 172770636-172770636 on Build GRCh38
rs13176979 Chr.5: 172770546-172770546 on Build GRCh38
rs34507926 Chr.5: 172770578-172770578 on Build GRCh38

View Full Product Details