Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46921142-46921403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP35
Entrez Gene ID
2909
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
UniGene
Hs.509447
Gene Chromosome Location
Chr.19: 46861002-47005077 on Build GRCh38
Transcripts
NM_004491.4 XM_011526873.2 XM_017026714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6085252 c.2503C>G p.L835V Chr.19: 46921178-46921178 on Build GRCh38
6337650 c.2626G>T p.E876* Chr.19: 46921301-46921301 on Build GRCh38
5802076 c.2530delT p.F844fs*43 Chr.19: 46921205-46921205 on Build GRCh38
3362926 c.2632C>A p.Q878K Chr.19: 46921307-46921307 on Build GRCh38
4482564 c.2620C>T p.L874F Chr.19: 46921295-46921295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758937587 Chr.19: 46921339-46921339 on Build GRCh38
rs753640209 Chr.19: 46921356-46921356 on Build GRCh38
rs377617202 Chr.19: 46921316-46921316 on Build GRCh38
rs542766066 Chr.19: 46921181-46921181 on Build GRCh38
rs755129329 Chr.19: 46921294-46921294 on Build GRCh38
rs188397648 Chr.19: 46921240-46921240 on Build GRCh38
rs991596100 Chr.19: 46921273-46921273 on Build GRCh38
rs201289408 Chr.19: 46921348-46921348 on Build GRCh38
rs1031456574 Chr.19: 46921278-46921278 on Build GRCh38
rs562764311 Chr.19: 46921224-46921224 on Build GRCh38
rs763897284 Chr.19: 46921248-46921248 on Build GRCh38
rs773808872 Chr.19: 46921189-46921189 on Build GRCh38
rs201565056 Chr.19: 46921349-46921349 on Build GRCh38
rs767101231 Chr.19: 46921343-46921343 on Build GRCh38
rs757142299 Chr.19: 46921268-46921268 on Build GRCh38
rs914262178 Chr.19: 46921378-46921378 on Build GRCh38
rs767071078 Chr.19: 46921229-46921229 on Build GRCh38
rs773747975 Chr.19: 46921219-46921219 on Build GRCh38
rs373836467 Chr.19: 46921334-46921334 on Build GRCh38
rs376792949 Chr.19: 46921170-46921170 on Build GRCh38
rs755879808 Chr.19: 46921241-46921241 on Build GRCh38
rs199618071 Chr.19: 46921346-46921346 on Build GRCh38
rs369352248 Chr.19: 46921288-46921288 on Build GRCh38
rs765848742 Chr.19: 46921206-46921206 on Build GRCh38
rs763587380 Chr.19: 46921205-46921205 on Build GRCh38
rs781546300 Chr.19: 46921298-46921298 on Build GRCh38
rs373792979 Chr.19: 46921303-46921303 on Build GRCh38
rs201791259 Chr.19: 46921367-46921367 on Build GRCh38
rs373804249 Chr.19: 46921366-46921366 on Build GRCh38
rs780026082 Chr.19: 46921169-46921169 on Build GRCh38
rs752386164 Chr.19: 46921237-46921237 on Build GRCh38
rs780120522 Chr.19: 46921286-46921286 on Build GRCh38
rs776981898 Chr.19: 46921176-46921176 on Build GRCh38
rs748496375 Chr.19: 46921178-46921178 on Build GRCh38
rs770244885 Chr.19: 46921315-46921315 on Build GRCh38
rs369651196 Chr.19: 46921321-46921321 on Build GRCh38
rs753731428 Chr.19: 46921260-46921260 on Build GRCh38
rs775778203 Chr.19: 46921331-46921331 on Build GRCh38

View Full Product Details