Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 74290124-74290377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD4
Entrez Gene ID
5826
Gene Name
ATP binding cassette subfamily D member 4
Gene Aliases
ABC41, EST352188, MAHCJ, P70R, P79R, PMP69, PXMP1L
UniGene
Hs.94395
Gene Chromosome Location
Chr.14: 74285277-74303064 on Build GRCh38
Transcripts
NM_005050.3 NR_003256.2 XM_005267939.3 XM_005267940.3 XM_005267941.4 XM_005267942.4 XM_005267946.3 XM_005267949.3 XM_006720223.2 XM_011537041.2 XM_017021531.1 XM_017021532.1 XM_017021533.1 XM_017021534.1 XM_017021535.1 XM_017021536.1 XM_017021537.1 XM_017021538.1 XM_017021539.1 XM_017021540.1 XM_017021541.1 XM_017021542.1 XM_017021543.1 XM_017021544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371046 c.1322C>T p.T441I Chr.14: 74290296-74290296 on Build GRCh38
1707598 c.1286C>T p.S429F Chr.14: 74290332-74290332 on Build GRCh38
1640152 c.1279A>G p.K427E Chr.14: 74290339-74290339 on Build GRCh38
957740 c.1287C>A p.S429S Chr.14: 74290331-74290331 on Build GRCh38
6317287 c.1263C>A p.G421G Chr.14: 74290355-74290355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768330414 Chr.14: 74290344-74290344 on Build GRCh38
rs201691861 Chr.14: 74290324-74290324 on Build GRCh38
rs11845976 Chr.14: 74290316-74290316 on Build GRCh38
rs745414252 Chr.14: 74290323-74290323 on Build GRCh38
rs370799934 Chr.14: 74290165-74290165 on Build GRCh38
rs139651889 Chr.14: 74290290-74290290 on Build GRCh38
rs746721131 Chr.14: 74290282-74290282 on Build GRCh38
rs751643150 Chr.14: 74290153-74290153 on Build GRCh38
rs770426895 Chr.14: 74290251-74290251 on Build GRCh38
rs199986349 Chr.14: 74290293-74290293 on Build GRCh38
rs750579449 Chr.14: 74290295-74290295 on Build GRCh38
rs767691217 Chr.14: 74290294-74290294 on Build GRCh38
rs993168511 Chr.14: 74290348-74290348 on Build GRCh38
rs1025552237 Chr.14: 74290225-74290225 on Build GRCh38
rs539308521 Chr.14: 74290203-74290203 on Build GRCh38
rs749042883 Chr.14: 74290328-74290328 on Build GRCh38
rs200761248 Chr.14: 74290302-74290302 on Build GRCh38
rs112493282 Chr.14: 74290282-74290282 on Build GRCh38
rs757424556 Chr.14: 74290160-74290160 on Build GRCh38
rs371381217 Chr.14: 74290247-74290247 on Build GRCh38
rs1043426774 Chr.14: 74290232-74290232 on Build GRCh38
rs200099391 Chr.14: 74290313-74290313 on Build GRCh38
rs368872131 Chr.14: 74290244-74290244 on Build GRCh38
rs530831051 Chr.14: 74290325-74290325 on Build GRCh38
rs910782736 Chr.14: 74290216-74290216 on Build GRCh38
rs776179662 Chr.14: 74290259-74290259 on Build GRCh38
rs769659682 Chr.14: 74290275-74290275 on Build GRCh38
rs756742956 Chr.14: 74290169-74290169 on Build GRCh38
rs199678602 Chr.14: 74290283-74290283 on Build GRCh38
rs117246905 Chr.14: 74290264-74290264 on Build GRCh38
rs780650462 Chr.14: 74290321-74290321 on Build GRCh38
rs1016529457 Chr.14: 74290188-74290188 on Build GRCh38
rs777309943 Chr.14: 74290249-74290249 on Build GRCh38
rs755309933 Chr.14: 74290307-74290307 on Build GRCh38
rs905092359 Chr.14: 74290296-74290296 on Build GRCh38
rs945270666 Chr.14: 74290335-74290335 on Build GRCh38
rs781736433 Chr.14: 74290161-74290161 on Build GRCh38
rs1026011443 Chr.14: 74290357-74290357 on Build GRCh38
rs567059239 Chr.14: 74290186-74290186 on Build GRCh38
rs746653976 Chr.14: 74290315-74290315 on Build GRCh38
rs774344504 Chr.14: 74290349-74290349 on Build GRCh38
rs780439786 Chr.14: 74290243-74290243 on Build GRCh38
rs971802263 Chr.14: 74290163-74290163 on Build GRCh38
rs954878832 Chr.14: 74290183-74290183 on Build GRCh38
rs201586690 Chr.14: 74290301-74290301 on Build GRCh38
rs557832539 Chr.14: 74290221-74290221 on Build GRCh38
rs537177278 Chr.14: 74290231-74290231 on Build GRCh38
rs544338934 Chr.14: 74290230-74290230 on Build GRCh38
rs774718245 Chr.14: 74290292-74290292 on Build GRCh38

View Full Product Details