Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120659725-120659990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP1
Entrez Gene ID
9478
Gene Name
calcium binding protein 1
Gene Aliases
CALBRAIN, HCALB_BR
UniGene
Hs.458482
Gene Chromosome Location
Chr.12: 120640552-120680869 on Build GRCh38
Transcripts
NM_001033677.1 NM_004276.4 NM_031205.3 XM_017020235.1 XM_017020236.1 XM_017020237.1 XM_017020238.1 XM_017020239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080942 c.682G>A p.E228K Chr.12: 120659905-120659905 on Build GRCh38
5953736 c.226-2A>G p.? Chr.12: 120659876-120659876 on Build GRCh38
7080943 c.253G>A p.E85K Chr.12: 120659905-120659905 on Build GRCh38
5583347 c.655-1G>A p.? Chr.12: 120659877-120659877 on Build GRCh38
5583348 c.226-1G>A p.? Chr.12: 120659877-120659877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767995821 Chr.12: 120659880-120659880 on Build GRCh38
rs994754471 Chr.12: 120659884-120659884 on Build GRCh38
rs999514465 Chr.12: 120659825-120659825 on Build GRCh38
rs3999390 Chr.12: 120659855-120659857 on Build GRCh38
rs7302002 Chr.12: 120659770-120659770 on Build GRCh38
rs754141707 Chr.12: 120659928-120659928 on Build GRCh38
rs529584 Chr.12: 120659819-120659819 on Build GRCh38
rs936321984 Chr.12: 120659798-120659798 on Build GRCh38
rs764851886 Chr.12: 120659857-120659857 on Build GRCh38
rs200270630 Chr.12: 120659932-120659932 on Build GRCh38
rs757394431 Chr.12: 120659929-120659929 on Build GRCh38
rs772135108 Chr.12: 120659840-120659840 on Build GRCh38
rs765714651 Chr.12: 120659847-120659847 on Build GRCh38
rs1011871988 Chr.12: 120659788-120659788 on Build GRCh38
rs967534394 Chr.12: 120659966-120659966 on Build GRCh38
rs142689476 Chr.12: 120659846-120659846 on Build GRCh38
rs753071042 Chr.12: 120659919-120659919 on Build GRCh38
rs764251003 Chr.12: 120659924-120659924 on Build GRCh38
rs3850511 Chr.12: 120659837-120659837 on Build GRCh38
rs764424777 Chr.12: 120659769-120659769 on Build GRCh38
rs772676449 Chr.12: 120659862-120659862 on Build GRCh38
rs906339654 Chr.12: 120659844-120659844 on Build GRCh38
rs369919458 Chr.12: 120659855-120659855 on Build GRCh38
rs373172687 Chr.12: 120659921-120659921 on Build GRCh38
rs3850512 Chr.12: 120659799-120659799 on Build GRCh38
rs977878459 Chr.12: 120659968-120659968 on Build GRCh38
rs961713710 Chr.12: 120659951-120659951 on Build GRCh38
rs1029540829 Chr.12: 120659889-120659889 on Build GRCh38
rs540769449 Chr.12: 120659813-120659813 on Build GRCh38
rs199748636 Chr.12: 120659833-120659833 on Build GRCh38
rs760101630 Chr.12: 120659863-120659863 on Build GRCh38
rs746832452 Chr.12: 120659954-120659954 on Build GRCh38
rs780072226 Chr.12: 120659952-120659952 on Build GRCh38
rs758425204 Chr.12: 120659943-120659943 on Build GRCh38
rs765949327 Chr.12: 120659839-120659839 on Build GRCh38
rs895029112 Chr.12: 120659804-120659804 on Build GRCh38
rs562814022 Chr.12: 120659849-120659849 on Build GRCh38
rs377611119 Chr.12: 120659956-120659956 on Build GRCh38
rs374211681 Chr.12: 120659848-120659848 on Build GRCh38
rs745932393 Chr.12: 120659935-120659935 on Build GRCh38
rs3999406 Chr.12: 120659856-120659858 on Build GRCh38

View Full Product Details