Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12044405-12044664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101928464
Entrez Gene ID
101928464
Gene Name
uncharacterized LOC101928464
Gene Aliases
-
UniGene
Hs.631643
Gene Chromosome Location
Chr.19: 11987617-12046275 on Build GRCh38
Transcripts
NR_134928.1 NR_134929.1 NM_001080404.2 XM_017027195.1 XM_017027196.1

Gene Symbol
ZNF878
Entrez Gene ID
729747
Gene Name
zinc finger protein 878
Gene Aliases
-
UniGene
Hs.704229
Gene Chromosome Location
Chr.19: 12043647-12052967 on Build GRCh38
Transcripts
NR_134928.1 NR_134929.1 NM_001080404.2 XM_017027195.1 XM_017027196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256130 c.1120_1121insCA p.R374fs*20 Chr.19: 12044421-12044422 on Build GRCh38
6833728 c.913C>T p.H305Y Chr.19: 12044488-12044488 on Build GRCh38
4511835 c.1022C>T p.S341F Chr.19: 12044520-12044520 on Build GRCh38
5500069 c.1026C>T p.V342V Chr.19: 12044516-12044516 on Build GRCh38
2815173 c.1121G>A p.R374H Chr.19: 12044421-12044421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768594889 Chr.19: 12044472-12044472 on Build GRCh38
rs566788694 Chr.19: 12044537-12044537 on Build GRCh38
rs375946184 Chr.19: 12044558-12044558 on Build GRCh38
rs370088952 Chr.19: 12044607-12044607 on Build GRCh38
rs200674509 Chr.19: 12044628-12044628 on Build GRCh38
rs753992039 Chr.19: 12044527-12044527 on Build GRCh38
rs377120157 Chr.19: 12044608-12044608 on Build GRCh38
rs746977680 Chr.19: 12044566-12044566 on Build GRCh38
rs921933128 Chr.19: 12044456-12044456 on Build GRCh38
rs778015043 Chr.19: 12044431-12044431 on Build GRCh38
rs768892496 Chr.19: 12044629-12044629 on Build GRCh38
rs749955533 Chr.19: 12044491-12044491 on Build GRCh38
rs745975070 Chr.19: 12044539-12044539 on Build GRCh38
rs756419765 Chr.19: 12044522-12044522 on Build GRCh38
rs200897750 Chr.19: 12044440-12044440 on Build GRCh38
rs373603905 Chr.19: 12044553-12044553 on Build GRCh38
rs776386296 Chr.19: 12044473-12044473 on Build GRCh38
rs369389389 Chr.19: 12044524-12044524 on Build GRCh38
rs776584874 Chr.19: 12044605-12044605 on Build GRCh38
rs75719391 Chr.19: 12044505-12044505 on Build GRCh38
rs975131554 Chr.19: 12044445-12044445 on Build GRCh38
rs753104457 Chr.19: 12044618-12044618 on Build GRCh38
rs749732053 Chr.19: 12044475-12044475 on Build GRCh38
rs760079798 Chr.19: 12044494-12044494 on Build GRCh38
rs939513690 Chr.19: 12044541-12044541 on Build GRCh38
rs745760062 Chr.19: 12044453-12044453 on Build GRCh38
rs767315864 Chr.19: 12044552-12044553 on Build GRCh38
rs770956433 Chr.19: 12044433-12044433 on Build GRCh38
rs779047914 Chr.19: 12044534-12044534 on Build GRCh38
rs939827523 Chr.19: 12044575-12044575 on Build GRCh38
rs749532414 Chr.19: 12044432-12044432 on Build GRCh38
rs371291916 Chr.19: 12044640-12044640 on Build GRCh38
rs201898665 Chr.19: 12044617-12044617 on Build GRCh38
rs761566734 Chr.19: 12044489-12044489 on Build GRCh38
rs771970824 Chr.19: 12044467-12044467 on Build GRCh38
rs757483478 Chr.19: 12044528-12044528 on Build GRCh38
rs751497019 Chr.19: 12044449-12044455 on Build GRCh38
rs760438433 Chr.19: 12044471-12044471 on Build GRCh38
rs757237751 Chr.19: 12044484-12044485 on Build GRCh38
rs762416726 Chr.19: 12044612-12044612 on Build GRCh38
rs768501176 Chr.19: 12044577-12044577 on Build GRCh38
rs533783513 Chr.19: 12044506-12044506 on Build GRCh38
rs372424657 Chr.19: 12044490-12044490 on Build GRCh38
rs780291735 Chr.19: 12044469-12044469 on Build GRCh38
rs878902321 Chr.19: 12044544-12044544 on Build GRCh38

View Full Product Details