Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53498510-53498735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
UniGene
Hs.380897
Gene Chromosome Location
Chr.16: 53491028-53504411 on Build GRCh38
Transcripts
NM_001012398.2 NM_001308325.1 NM_022476.3 XM_005256095.4 XM_005256096.4 XM_005256097.4 XM_005256098.4 XM_017023564.1 XM_017023565.1 XM_017023566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1679091 c.43C>T p.R15* Chr.16: 53498596-53498596 on Build GRCh38
6481553 c.97C>T p.R33* Chr.16: 53498542-53498542 on Build GRCh38
242893 c.76G>C p.D26H Chr.16: 53498563-53498563 on Build GRCh38
3510253 c.125C>T p.S42F Chr.16: 53498514-53498514 on Build GRCh38
971479 c.98G>A p.R33Q Chr.16: 53498541-53498541 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780618713 Chr.16: 53498623-53498624 on Build GRCh38
rs747266345 Chr.16: 53498649-53498649 on Build GRCh38
rs946558367 Chr.16: 53498664-53498664 on Build GRCh38
rs774604373 Chr.16: 53498600-53498600 on Build GRCh38
rs776539900 Chr.16: 53498560-53498560 on Build GRCh38
rs775441301 Chr.16: 53498579-53498579 on Build GRCh38
rs766797721 Chr.16: 53498621-53498621 on Build GRCh38
rs377629353 Chr.16: 53498620-53498620 on Build GRCh38
rs535466451 Chr.16: 53498689-53498689 on Build GRCh38
rs754972347 Chr.16: 53498652-53498652 on Build GRCh38
rs114541961 Chr.16: 53498661-53498661 on Build GRCh38
rs764155782 Chr.16: 53498595-53498595 on Build GRCh38
rs748620330 Chr.16: 53498697-53498697 on Build GRCh38
rs919044600 Chr.16: 53498581-53498581 on Build GRCh38
rs758843118 Chr.16: 53498566-53498567 on Build GRCh38
rs973534183 Chr.16: 53498566-53498566 on Build GRCh38
rs937647310 Chr.16: 53498632-53498632 on Build GRCh38
rs141560947 Chr.16: 53498541-53498541 on Build GRCh38
rs767686595 Chr.16: 53498615-53498615 on Build GRCh38
rs576136143 Chr.16: 53498669-53498669 on Build GRCh38
rs61734118 Chr.16: 53498576-53498576 on Build GRCh38
rs958720736 Chr.16: 53498667-53498667 on Build GRCh38
rs750775020 Chr.16: 53498618-53498618 on Build GRCh38
rs373543882 Chr.16: 53498607-53498607 on Build GRCh38
rs757855483 Chr.16: 53498537-53498537 on Build GRCh38
rs150363429 Chr.16: 53498656-53498656 on Build GRCh38
rs770900884 Chr.16: 53498547-53498547 on Build GRCh38
rs747376580 Chr.16: 53498648-53498650 on Build GRCh38
rs140403391 Chr.16: 53498588-53498588 on Build GRCh38
rs745639499 Chr.16: 53498561-53498561 on Build GRCh38

View Full Product Details