Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 13305336-13305609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTY
Entrez Gene ID
7404
Gene Name
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases
KDM6AL, UTY1
UniGene
Hs.115277
Gene Chromosome Location
Chr.Y: 13230770-13480670 on Build GRCh38
Transcripts
NM_001258249.1 NM_001258250.1 NM_001258251.1 NM_001258252.1 NM_001258253.1 NM_001258254.1 NM_001258255.1 NM_001258256.1 NM_001258257.1 NM_001258258.1 NM_001258259.1 NM_001258260.1 NM_001258261.1 NM_001258262.1 NM_001258263.1 NM_001258264.1 NM_001258265.1 NM_001258266.1 NM_001258267.1 NM_001258268.1 NM_001258269.1 NM_001258270.1 NM_007125.4 NM_182659.1 NR_047596.1 NR_047597.1 NR_047598.1 NR_047599.1 NR_047600.1 NR_047601.1 NR_047602.1 NR_047603.1 NR_047604.1 NR_047605.1 NR_047606.1 NR_047607.1 NR_047608.1 NR_047609.1 NR_047610.1 NR_047611.1 NR_047612.1 NR_047613.1 NR_047614.1 NR_047615.1 NR_047616.1 NR_047617.1 NR_047618.1 NR_047619.1 NR_047620.1 NR_047621.1 NR_047622.1 NR_047623.1 NR_047624.1 NR_047625.1 NR_047626.1 NR_047627.1 NR_047628.1 NR_047629.1 NR_047630.1 NR_047631.1 NR_047632.1 NR_047633.1 NR_047634.1 NR_047635.1 NR_047636.1 NR_047637.1 NR_047638.1 NR_047639.1 NR_047640.1 NR_047641.1 NR_047642.1 NR_047643.1 NR_047644.1 NR_047645.1 NR_047646.1 NR_047647.1 XM_005262518.3 XM_006724875.3 XM_011531441.2 XM_011531442.2 XM_011531443.2 XM_011531445.2 XM_011531446.2 XM_011531447.2 XM_011531448.2 XM_011531449.2 XM_011531450.2 XM_011531451.2 XM_011531452.2 XM_011531453.2 XM_011531454.2 XM_011531455.2 XM_011531457.2 XM_011531458.2 XM_011531459.2 XM_017030066.1 XM_017030067.1 XM_017030068.1 XM_017030069.1 XM_017030070.1 XM_017030071.1 XM_017030073.1 XM_017030074.1

SNP Detail

SNP ID Chromosome Location
rs925880782 Chr.Y: 13305477-13305477 on Build GRCh38
rs760465060 Chr.Y: 13305365-13305365 on Build GRCh38
rs760022192 Chr.Y: 13305371-13305371 on Build GRCh38
rs763288826 Chr.Y: 13305463-13305463 on Build GRCh38
rs767515011 Chr.Y: 13305500-13305500 on Build GRCh38
rs750992458 Chr.Y: 13305501-13305501 on Build GRCh38
rs754119788 Chr.Y: 13305582-13305582 on Build GRCh38
rs776280411 Chr.Y: 13305461-13305461 on Build GRCh38
rs761890490 Chr.Y: 13305489-13305489 on Build GRCh38
rs780515287 Chr.Y: 13305551-13305551 on Build GRCh38
rs200431840 Chr.Y: 13305480-13305480 on Build GRCh38
rs776828569 Chr.Y: 13305357-13305357 on Build GRCh38
rs370331284 Chr.Y: 13305508-13305508 on Build GRCh38
rs1014966916 Chr.Y: 13305559-13305559 on Build GRCh38
rs760836256 Chr.Y: 13305500-13305500 on Build GRCh38
rs764195833 Chr.Y: 13305469-13305469 on Build GRCh38

View Full Product Details