Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68655120-68655359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B15
Entrez Gene ID
7366
Gene Name
UDP glucuronosyltransferase family 2 member B15
Gene Aliases
HLUG4, UDPGT 2B8, UDPGT2B15, UDPGTH3, UGT2B8
UniGene
Hs.150207
Gene Chromosome Location
Chr.4: 68646597-68670776 on Build GRCh38
Transcripts
NM_001076.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6100549 c.1053T>A p.N351K Chr.4: 68655135-68655135 on Build GRCh38
3605115 c.1014G>A p.W338* Chr.4: 68655174-68655174 on Build GRCh38
5995550 c.1063T>C p.Y355H Chr.4: 68655125-68655125 on Build GRCh38
5653998 c.1006G>T p.V336F Chr.4: 68655182-68655182 on Build GRCh38
7140733 c.1041T>G p.T347T Chr.4: 68655147-68655147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779446832 Chr.4: 68655164-68655164 on Build GRCh38
rs903813091 Chr.4: 68655237-68655237 on Build GRCh38
rs759369248 Chr.4: 68655186-68655186 on Build GRCh38
rs771566387 Chr.4: 68655171-68655171 on Build GRCh38
rs369116334 Chr.4: 68655216-68655216 on Build GRCh38
rs532134840 Chr.4: 68655266-68655269 on Build GRCh38
rs539442094 Chr.4: 68655222-68655222 on Build GRCh38
rs377477047 Chr.4: 68655193-68655193 on Build GRCh38
rs1023991444 Chr.4: 68655235-68655235 on Build GRCh38
rs1012135062 Chr.4: 68655231-68655231 on Build GRCh38
rs765125890 Chr.4: 68655187-68655187 on Build GRCh38
rs368303965 Chr.4: 68655205-68655205 on Build GRCh38
rs975654741 Chr.4: 68655317-68655317 on Build GRCh38
rs757524217 Chr.4: 68655234-68655236 on Build GRCh38
rs142948482 Chr.4: 68655149-68655149 on Build GRCh38
rs1000828723 Chr.4: 68655244-68655244 on Build GRCh38
rs770860129 Chr.4: 68655179-68655179 on Build GRCh38
rs920181955 Chr.4: 68655318-68655318 on Build GRCh38
rs780334832 Chr.4: 68655298-68655300 on Build GRCh38
rs964581412 Chr.4: 68655300-68655300 on Build GRCh38
rs147708073 Chr.4: 68655159-68655159 on Build GRCh38
rs755506837 Chr.4: 68655161-68655161 on Build GRCh38
rs747665883 Chr.4: 68655165-68655165 on Build GRCh38
rs766584868 Chr.4: 68655233-68655233 on Build GRCh38
rs756388748 Chr.4: 68655230-68655230 on Build GRCh38
rs746589344 Chr.4: 68655177-68655177 on Build GRCh38
rs776623377 Chr.4: 68655183-68655183 on Build GRCh38
rs773027308 Chr.4: 68655174-68655174 on Build GRCh38
rs761819987 Chr.4: 68655197-68655197 on Build GRCh38
rs986876084 Chr.4: 68655295-68655295 on Build GRCh38
rs1031002275 Chr.4: 68655252-68655252 on Build GRCh38

View Full Product Details