Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232748942-232749197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5862015 c.199T>C p.F67L Chr.2: 232749014-232749014 on Build GRCh38
3407669 c.267+2T>C p.? Chr.2: 232749084-232749084 on Build GRCh38
1591947 c.196G>T p.E66* Chr.2: 232749011-232749011 on Build GRCh38
3579123 c.201T>C p.F67F Chr.2: 232749016-232749016 on Build GRCh38
5728130 c.183C>T p.D61D Chr.2: 232748998-232748998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777409563 Chr.2: 232748996-232748996 on Build GRCh38
rs747850665 Chr.2: 232748979-232748979 on Build GRCh38
rs533112093 Chr.2: 232749146-232749146 on Build GRCh38
rs368053818 Chr.2: 232749120-232749120 on Build GRCh38
rs557364792 Chr.2: 232749096-232749096 on Build GRCh38
rs766911895 Chr.2: 232749079-232749079 on Build GRCh38
rs754373854 Chr.2: 232749085-232749085 on Build GRCh38
rs906106304 Chr.2: 232749101-232749101 on Build GRCh38
rs759509725 Chr.2: 232749019-232749019 on Build GRCh38
rs755420281 Chr.2: 232749090-232749090 on Build GRCh38
rs143370656 Chr.2: 232749004-232749004 on Build GRCh38
rs770600253 Chr.2: 232749118-232749118 on Build GRCh38
rs769730506 Chr.2: 232749020-232749020 on Build GRCh38
rs770953286 Chr.2: 232749007-232749007 on Build GRCh38
rs754522961 Chr.2: 232749112-232749113 on Build GRCh38
rs377476685 Chr.2: 232749065-232749065 on Build GRCh38
rs192688643 Chr.2: 232749103-232749103 on Build GRCh38
rs760647183 Chr.2: 232749073-232749073 on Build GRCh38
rs201232635 Chr.2: 232749104-232749104 on Build GRCh38
rs750572791 Chr.2: 232749064-232749064 on Build GRCh38
rs924924038 Chr.2: 232748992-232748992 on Build GRCh38
rs886579047 Chr.2: 232749134-232749134 on Build GRCh38
rs370317126 Chr.2: 232749099-232749099 on Build GRCh38
rs573169813 Chr.2: 232748987-232748987 on Build GRCh38
rs762096898 Chr.2: 232749030-232749030 on Build GRCh38
rs747570430 Chr.2: 232749102-232749102 on Build GRCh38
rs374565166 Chr.2: 232749097-232749097 on Build GRCh38
rs1003983744 Chr.2: 232749160-232749160 on Build GRCh38
rs916889768 Chr.2: 232749025-232749025 on Build GRCh38
rs371199763 Chr.2: 232749023-232749023 on Build GRCh38
rs374831276 Chr.2: 232749056-232749056 on Build GRCh38
rs954688997 Chr.2: 232749165-232749165 on Build GRCh38
rs776682435 Chr.2: 232749017-232749017 on Build GRCh38

View Full Product Details