Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8256154-8256415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFAS
Entrez Gene ID
5198
Gene Name
phosphoribosylformylglycinamidine synthase
Gene Aliases
FGAMS, FGAR-AT, FGARAT, PURL
UniGene
Hs.573976
Gene Chromosome Location
Chr.17: 8249255-8270491 on Build GRCh38
Transcripts
NM_012393.2 XM_006721546.2 XM_017024760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828911 c.692G>T p.R231L Chr.17: 8256278-8256278 on Build GRCh38
3937675 c.758C>G p.S253C Chr.17: 8256344-8256344 on Build GRCh38
6744199 c.783G>A p.S261S Chr.17: 8256369-8256369 on Build GRCh38
6744209 c.775G>T p.E259* Chr.17: 8256361-8256361 on Build GRCh38
6016477 c.774G>A p.Q258Q Chr.17: 8256360-8256360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921850040 Chr.17: 8256347-8256347 on Build GRCh38
rs373102792 Chr.17: 8256232-8256232 on Build GRCh38
rs576338195 Chr.17: 8256195-8256195 on Build GRCh38
rs944531383 Chr.17: 8256300-8256300 on Build GRCh38
rs930348518 Chr.17: 8256348-8256348 on Build GRCh38
rs769796370 Chr.17: 8256381-8256381 on Build GRCh38
rs778901080 Chr.17: 8256259-8256259 on Build GRCh38
rs371281535 Chr.17: 8256359-8256359 on Build GRCh38
rs770011828 Chr.17: 8256227-8256227 on Build GRCh38
rs750037224 Chr.17: 8256211-8256211 on Build GRCh38
rs543836971 Chr.17: 8256216-8256216 on Build GRCh38
rs774909629 Chr.17: 8256315-8256315 on Build GRCh38
rs201102078 Chr.17: 8256238-8256238 on Build GRCh38
rs746841927 Chr.17: 8256274-8256274 on Build GRCh38
rs746784891 Chr.17: 8256390-8256390 on Build GRCh38
rs779405841 Chr.17: 8256382-8256382 on Build GRCh38
rs970041387 Chr.17: 8256375-8256375 on Build GRCh38
rs368386929 Chr.17: 8256372-8256372 on Build GRCh38
rs753373433 Chr.17: 8256255-8256255 on Build GRCh38
rs1035560707 Chr.17: 8256222-8256222 on Build GRCh38
rs775947544 Chr.17: 8256341-8256341 on Build GRCh38
rs147784624 Chr.17: 8256349-8256349 on Build GRCh38
rs558943027 Chr.17: 8256267-8256267 on Build GRCh38
rs376283565 Chr.17: 8256218-8256218 on Build GRCh38
rs758372627 Chr.17: 8256268-8256268 on Build GRCh38
rs758231026 Chr.17: 8256378-8256378 on Build GRCh38
rs1023877639 Chr.17: 8256235-8256235 on Build GRCh38
rs1031119035 Chr.17: 8256258-8256258 on Build GRCh38
rs779097739 Chr.17: 8256256-8256257 on Build GRCh38
rs769958484 Chr.17: 8256277-8256277 on Build GRCh38
rs748315593 Chr.17: 8256226-8256226 on Build GRCh38
rs753116264 Chr.17: 8256247-8256247 on Build GRCh38
rs376893488 Chr.17: 8256256-8256256 on Build GRCh38
rs779760832 Chr.17: 8256272-8256272 on Build GRCh38
rs753496976 Chr.17: 8256362-8256362 on Build GRCh38
rs764590509 Chr.17: 8256369-8256369 on Build GRCh38
rs759892673 Chr.17: 8256335-8256335 on Build GRCh38
rs911697978 Chr.17: 8256278-8256278 on Build GRCh38
rs372662450 Chr.17: 8256252-8256252 on Build GRCh38
rs767714917 Chr.17: 8256245-8256245 on Build GRCh38
rs755786686 Chr.17: 8256248-8256248 on Build GRCh38
rs1048845299 Chr.17: 8256206-8256206 on Build GRCh38
rs376354399 Chr.17: 8256364-8256364 on Build GRCh38
rs541236125 Chr.17: 8256306-8256306 on Build GRCh38
rs200924377 Chr.17: 8256273-8256273 on Build GRCh38
rs772401956 Chr.17: 8256336-8256337 on Build GRCh38
rs377030754 Chr.17: 8256197-8256197 on Build GRCh38
rs888167798 Chr.17: 8256261-8256261 on Build GRCh38
rs762966351 Chr.17: 8256234-8256234 on Build GRCh38
rs759941027 Chr.17: 8256239-8256239 on Build GRCh38
rs768167913 Chr.17: 8256338-8256338 on Build GRCh38
rs555136594 Chr.17: 8256187-8256187 on Build GRCh38
rs770985519 Chr.17: 8256308-8256308 on Build GRCh38
rs763805801 Chr.17: 8256360-8256360 on Build GRCh38
rs745985584 Chr.17: 8256335-8256338 on Build GRCh38
rs144281287 Chr.17: 8256307-8256307 on Build GRCh38
rs926007194 Chr.17: 8256210-8256210 on Build GRCh38

View Full Product Details