Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39779113-39779360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF5
Entrez Gene ID
150084
Gene Name
immunoglobulin superfamily member 5
Gene Aliases
GSF5, JAM4
UniGene
Hs.422120
Gene Chromosome Location
Chr.21: 39725838-39802096 on Build GRCh38
Transcripts
NM_001080444.1 XM_011529472.1 XM_011529473.1 XM_017028279.1 XM_017028280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5392900 c.817G>A p.G273S Chr.21: 39779188-39779188 on Build GRCh38
1030780 c.856C>T p.R286C Chr.21: 39779227-39779227 on Build GRCh38
3963984 c.745G>C p.G249R Chr.21: 39779116-39779116 on Build GRCh38
1030779 c.855A>T p.I285I Chr.21: 39779226-39779226 on Build GRCh38
1030778 c.834G>A p.T278T Chr.21: 39779205-39779205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762179531 Chr.21: 39779239-39779240 on Build GRCh38
rs752381457 Chr.21: 39779148-39779148 on Build GRCh38
rs764468617 Chr.21: 39779304-39779304 on Build GRCh38
rs1008080471 Chr.21: 39779211-39779211 on Build GRCh38
rs143191361 Chr.21: 39779236-39779236 on Build GRCh38
rs142136474 Chr.21: 39779195-39779195 on Build GRCh38
rs759460658 Chr.21: 39779221-39779221 on Build GRCh38
rs775043014 Chr.21: 39779228-39779228 on Build GRCh38
rs763627176 Chr.21: 39779162-39779162 on Build GRCh38
rs150645419 Chr.21: 39779249-39779249 on Build GRCh38
rs368677603 Chr.21: 39779156-39779156 on Build GRCh38
rs78244022 Chr.21: 39779276-39779276 on Build GRCh38
rs963359781 Chr.21: 39779272-39779272 on Build GRCh38
rs374675570 Chr.21: 39779261-39779261 on Build GRCh38
rs573576042 Chr.21: 39779241-39779241 on Build GRCh38
rs200284688 Chr.21: 39779208-39779208 on Build GRCh38
rs761204033 Chr.21: 39779264-39779264 on Build GRCh38
rs764698988 Chr.21: 39779287-39779287 on Build GRCh38
rs745784705 Chr.21: 39779253-39779253 on Build GRCh38
rs779112202 Chr.21: 39779311-39779311 on Build GRCh38
rs926019670 Chr.21: 39779227-39779227 on Build GRCh38
rs865947636 Chr.21: 39779159-39779159 on Build GRCh38
rs758362558 Chr.21: 39779315-39779315 on Build GRCh38
rs112596313 Chr.21: 39779165-39779165 on Build GRCh38
rs569416566 Chr.21: 39779205-39779205 on Build GRCh38
rs868811340 Chr.21: 39779150-39779150 on Build GRCh38
rs779059120 Chr.21: 39779251-39779251 on Build GRCh38
rs772649631 Chr.21: 39779249-39779251 on Build GRCh38
rs753107016 Chr.21: 39779298-39779298 on Build GRCh38
rs771895470 Chr.21: 39779222-39779222 on Build GRCh38
rs773501685 Chr.21: 39779272-39779273 on Build GRCh38
rs200406404 Chr.21: 39779234-39779234 on Build GRCh38
rs530090000 Chr.21: 39779227-39779228 on Build GRCh38
rs779799102 Chr.21: 39779316-39779316 on Build GRCh38
rs143743565 Chr.21: 39779223-39779223 on Build GRCh38
rs758936711 Chr.21: 39779176-39779176 on Build GRCh38
rs778071541 Chr.21: 39779248-39779248 on Build GRCh38
rs957679347 Chr.21: 39779230-39779231 on Build GRCh38
rs372540912 Chr.21: 39779149-39779149 on Build GRCh38
rs757398362 Chr.21: 39779250-39779250 on Build GRCh38
rs556241853 Chr.21: 39779300-39779300 on Build GRCh38
rs369163594 Chr.21: 39779245-39779245 on Build GRCh38
rs774890521 Chr.21: 39779288-39779288 on Build GRCh38
rs760114420 Chr.21: 39779296-39779296 on Build GRCh38
rs780473003 Chr.21: 39779186-39779186 on Build GRCh38
rs185320685 Chr.21: 39779310-39779310 on Build GRCh38
rs555594699 Chr.21: 39779203-39779203 on Build GRCh38
rs192405201 Chr.21: 39779242-39779242 on Build GRCh38
rs774372928 Chr.21: 39779219-39779219 on Build GRCh38
rs760398900 Chr.21: 39779232-39779232 on Build GRCh38
rs201593108 Chr.21: 39779275-39779275 on Build GRCh38
rs749337072 Chr.21: 39779210-39779210 on Build GRCh38
rs371394692 Chr.21: 39779256-39779256 on Build GRCh38
rs1018090835 Chr.21: 39779224-39779224 on Build GRCh38
rs754107936 Chr.21: 39779306-39779306 on Build GRCh38
rs763614592 Chr.21: 39779233-39779233 on Build GRCh38
rs200941561 Chr.21: 39779207-39779207 on Build GRCh38
rs199672249 Chr.21: 39779246-39779246 on Build GRCh38
rs369881689 Chr.21: 39779262-39779262 on Build GRCh38
rs1051635966 Chr.21: 39779160-39779160 on Build GRCh38
rs927466034 Chr.21: 39779243-39779243 on Build GRCh38
rs768021550 Chr.21: 39779236-39779237 on Build GRCh38
rs779839131 Chr.21: 39779228-39779230 on Build GRCh38
rs770992693 Chr.21: 39779215-39779215 on Build GRCh38
rs768095817 Chr.21: 39779233-39779234 on Build GRCh38
rs371965247 Chr.21: 39779204-39779204 on Build GRCh38
rs536847797 Chr.21: 39779161-39779161 on Build GRCh38
rs148238696 Chr.21: 39779239-39779239 on Build GRCh38
rs898158475 Chr.21: 39779178-39779178 on Build GRCh38

View Full Product Details