Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4489069-4489310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52K1
Entrez Gene ID
390036
Gene Name
olfactory receptor family 52 subfamily K member 1
Gene Aliases
OR11-8
UniGene
Hs.553726
Gene Chromosome Location
Chr.11: 4488879-4489908 on Build GRCh38
Transcripts
NM_001005171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
542550 c.263G>T p.W88L Chr.11: 4489163-4489163 on Build GRCh38
4032857 c.356C>A p.A119D Chr.11: 4489256-4489256 on Build GRCh38
1353834 c.370C>T p.R124C Chr.11: 4489270-4489270 on Build GRCh38
322357 c.403A>G p.T135A Chr.11: 4489303-4489303 on Build GRCh38
4032858 c.405G>A p.T135T Chr.11: 4489305-4489305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964058879 Chr.11: 4489102-4489102 on Build GRCh38
rs777771146 Chr.11: 4489139-4489139 on Build GRCh38
rs767531406 Chr.11: 4489240-4489240 on Build GRCh38
rs1036628983 Chr.11: 4489241-4489241 on Build GRCh38
rs769595321 Chr.11: 4489108-4489108 on Build GRCh38
rs768344079 Chr.11: 4489182-4489182 on Build GRCh38
rs770839729 Chr.11: 4489100-4489100 on Build GRCh38
rs370066776 Chr.11: 4489206-4489206 on Build GRCh38
rs771712348 Chr.11: 4489279-4489279 on Build GRCh38
rs1008755874 Chr.11: 4489209-4489209 on Build GRCh38
rs781010424 Chr.11: 4489104-4489104 on Build GRCh38
rs779835487 Chr.11: 4489175-4489175 on Build GRCh38
rs748963848 Chr.11: 4489177-4489177 on Build GRCh38
rs758639587 Chr.11: 4489138-4489138 on Build GRCh38
rs141745573 Chr.11: 4489257-4489257 on Build GRCh38
rs757222438 Chr.11: 4489155-4489155 on Build GRCh38
rs368528167 Chr.11: 4489271-4489271 on Build GRCh38
rs765578382 Chr.11: 4489137-4489137 on Build GRCh38
rs138616789 Chr.11: 4489112-4489112 on Build GRCh38
rs745316833 Chr.11: 4489190-4489190 on Build GRCh38
rs147734448 Chr.11: 4489263-4489263 on Build GRCh38
rs751737693 Chr.11: 4489219-4489219 on Build GRCh38
rs145753082 Chr.11: 4489106-4489106 on Build GRCh38
rs147098497 Chr.11: 4489260-4489260 on Build GRCh38
rs763130739 Chr.11: 4489207-4489207 on Build GRCh38
rs200029336 Chr.11: 4489113-4489113 on Build GRCh38
rs201104279 Chr.11: 4489136-4489136 on Build GRCh38
rs142040619 Chr.11: 4489097-4489097 on Build GRCh38
rs537539173 Chr.11: 4489174-4489174 on Build GRCh38
rs147596013 Chr.11: 4489200-4489200 on Build GRCh38
rs139078347 Chr.11: 4489145-4489145 on Build GRCh38
rs376395151 Chr.11: 4489127-4489127 on Build GRCh38
rs757354444 Chr.11: 4489224-4489224 on Build GRCh38
rs568987040 Chr.11: 4489135-4489135 on Build GRCh38
rs568373516 Chr.11: 4489107-4489107 on Build GRCh38
rs771669265 Chr.11: 4489193-4489193 on Build GRCh38
rs771791857 Chr.11: 4489107-4489107 on Build GRCh38
rs772842963 Chr.11: 4489280-4489280 on Build GRCh38
rs774055555 Chr.11: 4489189-4489189 on Build GRCh38
rs754626592 Chr.11: 4489268-4489268 on Build GRCh38
rs747013101 Chr.11: 4489098-4489098 on Build GRCh38
rs777291681 Chr.11: 4489196-4489196 on Build GRCh38
rs746485765 Chr.11: 4489227-4489229 on Build GRCh38
rs940420928 Chr.11: 4489168-4489168 on Build GRCh38
rs890404112 Chr.11: 4489194-4489194 on Build GRCh38
rs764342505 Chr.11: 4489215-4489215 on Build GRCh38
rs897633516 Chr.11: 4489256-4489256 on Build GRCh38
rs773003076 Chr.11: 4489221-4489226 on Build GRCh38
rs781135063 Chr.11: 4489156-4489156 on Build GRCh38
rs745761061 Chr.11: 4489164-4489164 on Build GRCh38
rs765586746 Chr.11: 4489202-4489202 on Build GRCh38
rs200982565 Chr.11: 4489270-4489270 on Build GRCh38
rs202235407 Chr.11: 4489188-4489188 on Build GRCh38

View Full Product Details