Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6779815-6780084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO39
Entrez Gene ID
162517
Gene Name
F-box protein 39
Gene Aliases
CT144, Fbx39
UniGene
Hs.368364
Gene Chromosome Location
Chr.17: 6776184-6798929 on Build GRCh38
Transcripts
NM_153230.2 XM_011523697.1 XM_017024283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7215501 c.28C>T p.P10S Chr.17: 6779896-6779896 on Build GRCh38
2975072 c.6C>T p.D2D Chr.17: 6779874-6779874 on Build GRCh38
4167496 c.76C>T p.R26C Chr.17: 6779944-6779944 on Build GRCh38
1661822 c.131G>C p.R44T Chr.17: 6779999-6779999 on Build GRCh38
6465104 c.22A>T p.I8F Chr.17: 6779890-6779890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768687307 Chr.17: 6780060-6780060 on Build GRCh38
rs973567392 Chr.17: 6780036-6780036 on Build GRCh38
rs778419218 Chr.17: 6779902-6779902 on Build GRCh38
rs140674090 Chr.17: 6780038-6780038 on Build GRCh38
rs764332418 Chr.17: 6779848-6779848 on Build GRCh38
rs143419082 Chr.17: 6779872-6779872 on Build GRCh38
rs542378940 Chr.17: 6780019-6780019 on Build GRCh38
rs887378122 Chr.17: 6779916-6779916 on Build GRCh38
rs770428705 Chr.17: 6779899-6779899 on Build GRCh38
rs1006244933 Chr.17: 6779919-6779919 on Build GRCh38
rs376499600 Chr.17: 6780049-6780049 on Build GRCh38
rs150934591 Chr.17: 6779945-6779945 on Build GRCh38
rs61753286 Chr.17: 6779874-6779874 on Build GRCh38
rs780924976 Chr.17: 6779992-6779992 on Build GRCh38
rs749711767 Chr.17: 6779909-6779909 on Build GRCh38
rs369991116 Chr.17: 6780047-6780047 on Build GRCh38
rs762819097 Chr.17: 6780062-6780062 on Build GRCh38
rs146724926 Chr.17: 6779885-6779885 on Build GRCh38
rs368045455 Chr.17: 6779875-6779875 on Build GRCh38
rs774739920 Chr.17: 6779912-6779912 on Build GRCh38
rs751318642 Chr.17: 6779976-6779976 on Build GRCh38
rs755585129 Chr.17: 6779851-6779851 on Build GRCh38
rs749792809 Chr.17: 6780026-6780026 on Build GRCh38
rs755664200 Chr.17: 6780012-6780012 on Build GRCh38
rs747137501 Chr.17: 6780048-6780048 on Build GRCh38
rs368869386 Chr.17: 6779838-6779838 on Build GRCh38
rs201069945 Chr.17: 6779928-6779928 on Build GRCh38
rs4796555 Chr.17: 6779896-6779896 on Build GRCh38
rs576606128 Chr.17: 6779993-6779993 on Build GRCh38
rs964089461 Chr.17: 6779978-6779978 on Build GRCh38
rs759700825 Chr.17: 6779917-6779917 on Build GRCh38
rs776729788 Chr.17: 6780061-6780061 on Build GRCh38
rs778465960 Chr.17: 6780025-6780025 on Build GRCh38
rs766116765 Chr.17: 6779971-6779971 on Build GRCh38
rs146370414 Chr.17: 6779941-6779941 on Build GRCh38
rs61753143 Chr.17: 6779890-6779890 on Build GRCh38
rs1010770497 Chr.17: 6779871-6779871 on Build GRCh38
rs754599697 Chr.17: 6779982-6779982 on Build GRCh38
rs772227770 Chr.17: 6779925-6779925 on Build GRCh38
rs761755341 Chr.17: 6779859-6779859 on Build GRCh38
rs765096770 Chr.17: 6779863-6779863 on Build GRCh38
rs78983367 Chr.17: 6779929-6779929 on Build GRCh38
rs200419578 Chr.17: 6780037-6780037 on Build GRCh38

View Full Product Details