Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210440541-210440804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LANCL1
Entrez Gene ID
10314
Gene Name
LanC like 1
Gene Aliases
GPR69A, p40
UniGene
Hs.13351
Gene Chromosome Location
Chr.2: 210431249-210477618 on Build GRCh38
Transcripts
NM_001136574.1 NM_001136575.1 NM_006055.2 XM_005246243.2 NR_110604.1 NR_110605.1 NR_110606.1

Gene Symbol
LANCL1-AS1
Entrez Gene ID
102724820
Gene Name
LANCL1 antisense RNA 1
Gene Aliases
LANCLOT
UniGene
Hs.659232
Gene Chromosome Location
Chr.2: 210324712-210470333 on Build GRCh38
Transcripts
NM_001136574.1 NM_001136575.1 NM_006055.2 XM_005246243.2 NR_110604.1 NR_110605.1 NR_110606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508017 c.669A>G p.G223G Chr.2: 210440619-210440619 on Build GRCh38
3380020 c.599C>T p.T200M Chr.2: 210440689-210440689 on Build GRCh38
4090769 c.646G>A p.G216R Chr.2: 210440642-210440642 on Build GRCh38
1015854 c.597C>A p.F199L Chr.2: 210440691-210440691 on Build GRCh38
2715559 c.572A>G p.E191G Chr.2: 210440716-210440716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767399633 Chr.2: 210440760-210440760 on Build GRCh38
rs751262200 Chr.2: 210440640-210440640 on Build GRCh38
rs759633804 Chr.2: 210440738-210440738 on Build GRCh38
rs770467794 Chr.2: 210440741-210440741 on Build GRCh38
rs755668534 Chr.2: 210440662-210440662 on Build GRCh38
rs946713321 Chr.2: 210440729-210440729 on Build GRCh38
rs1015786979 Chr.2: 210440705-210440705 on Build GRCh38
rs1004027804 Chr.2: 210440701-210440701 on Build GRCh38
rs756051191 Chr.2: 210440777-210440777 on Build GRCh38
rs766513528 Chr.2: 210440609-210440609 on Build GRCh38
rs922464964 Chr.2: 210440606-210440606 on Build GRCh38
rs756995112 Chr.2: 210440758-210440758 on Build GRCh38
rs748440609 Chr.2: 210440684-210440684 on Build GRCh38
rs200838829 Chr.2: 210440626-210440626 on Build GRCh38
rs112032576 Chr.2: 210440709-210440709 on Build GRCh38
rs575545684 Chr.2: 210440586-210440586 on Build GRCh38
rs769921799 Chr.2: 210440658-210440658 on Build GRCh38
rs141178650 Chr.2: 210440689-210440689 on Build GRCh38
rs372217602 Chr.2: 210440631-210440631 on Build GRCh38
rs200162143 Chr.2: 210440678-210440678 on Build GRCh38
rs1002116199 Chr.2: 210440612-210440612 on Build GRCh38
rs753927270 Chr.2: 210440614-210440614 on Build GRCh38
rs748813502 Chr.2: 210440670-210440670 on Build GRCh38
rs768386883 Chr.2: 210440671-210440671 on Build GRCh38
rs750260340 Chr.2: 210440768-210440768 on Build GRCh38
rs1052567586 Chr.2: 210440667-210440667 on Build GRCh38
rs1045483802 Chr.2: 210440737-210440737 on Build GRCh38
rs746036722 Chr.2: 210440668-210440668 on Build GRCh38
rs773818246 Chr.2: 210440598-210440598 on Build GRCh38
rs765527139 Chr.2: 210440633-210440633 on Build GRCh38
rs752999125 Chr.2: 210440755-210440755 on Build GRCh38
rs757121112 Chr.2: 210440643-210440643 on Build GRCh38
rs766875108 Chr.2: 210440702-210440702 on Build GRCh38
rs553320476 Chr.2: 210440607-210440607 on Build GRCh38
rs112466780 Chr.2: 210440601-210440601 on Build GRCh38
rs141027852 Chr.2: 210440688-210440688 on Build GRCh38
rs761230098 Chr.2: 210440692-210440692 on Build GRCh38
rs780956260 Chr.2: 210440649-210440649 on Build GRCh38
rs776650427 Chr.2: 210440710-210440710 on Build GRCh38
rs765350994 Chr.2: 210440750-210440750 on Build GRCh38
rs767864226 Chr.2: 210440592-210440592 on Build GRCh38

View Full Product Details