Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94997907-94998180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4D
Entrez Gene ID
55693
Gene Name
lysine demethylase 4D
Gene Aliases
JMJD2D
UniGene
Hs.503598
Gene Chromosome Location
Chr.11: 94973681-94999514 on Build GRCh38
Transcripts
NM_018039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692503 c.545T>G p.F182C Chr.11: 94997917-94997917 on Build GRCh38
4694980 c.637T>C p.Y213H Chr.11: 94998009-94998009 on Build GRCh38
6045672 c.647C>T p.P216L Chr.11: 94998019-94998019 on Build GRCh38
1357983 c.771T>G p.N257K Chr.11: 94998143-94998143 on Build GRCh38
4842789 c.672A>G p.E224E Chr.11: 94998044-94998044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911469300 Chr.11: 94997951-94997951 on Build GRCh38
rs782491473 Chr.11: 94997988-94997988 on Build GRCh38
rs587703801 Chr.11: 94998149-94998149 on Build GRCh38
rs782381590 Chr.11: 94998046-94998046 on Build GRCh38
rs373019386 Chr.11: 94998160-94998160 on Build GRCh38
rs922940523 Chr.11: 94998081-94998081 on Build GRCh38
rs782086850 Chr.11: 94998121-94998121 on Build GRCh38
rs782677713 Chr.11: 94998042-94998042 on Build GRCh38
rs17680061 Chr.11: 94997948-94997948 on Build GRCh38
rs782199460 Chr.11: 94998159-94998159 on Build GRCh38
rs782075451 Chr.11: 94998125-94998125 on Build GRCh38
rs587721515 Chr.11: 94998027-94998027 on Build GRCh38
rs782231727 Chr.11: 94998069-94998069 on Build GRCh38
rs782771414 Chr.11: 94997941-94997941 on Build GRCh38
rs781979334 Chr.11: 94998108-94998108 on Build GRCh38
rs942961384 Chr.11: 94998036-94998036 on Build GRCh38
rs781929797 Chr.11: 94998078-94998078 on Build GRCh38
rs782495153 Chr.11: 94998155-94998155 on Build GRCh38
rs199767727 Chr.11: 94997972-94997972 on Build GRCh38
rs782310978 Chr.11: 94998073-94998073 on Build GRCh38
rs782536687 Chr.11: 94998037-94998037 on Build GRCh38
rs782662807 Chr.11: 94998143-94998143 on Build GRCh38
rs370480714 Chr.11: 94998147-94998147 on Build GRCh38
rs146449803 Chr.11: 94998157-94998157 on Build GRCh38
rs782429808 Chr.11: 94998107-94998107 on Build GRCh38
rs782605380 Chr.11: 94998062-94998062 on Build GRCh38
rs782796507 Chr.11: 94998127-94998127 on Build GRCh38
rs781908190 Chr.11: 94998128-94998128 on Build GRCh38
rs201415024 Chr.11: 94998079-94998079 on Build GRCh38
rs782245540 Chr.11: 94998045-94998045 on Build GRCh38
rs782499289 Chr.11: 94998142-94998142 on Build GRCh38
rs782575045 Chr.11: 94998011-94998011 on Build GRCh38
rs771482908 Chr.11: 94998152-94998152 on Build GRCh38
rs782156404 Chr.11: 94997936-94997936 on Build GRCh38

View Full Product Details