Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124405379-124405626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAT
Entrez Gene ID
4942
Gene Name
ornithine aminotransferase
Gene Aliases
GACR, HOGA, OATASE, OKT
UniGene
Hs.523332
Gene Chromosome Location
Chr.10: 124397303-124418976 on Build GRCh38
Transcripts
NM_000274.3 NM_001171814.1 NM_001322965.1 NM_001322966.1 NM_001322967.1 NM_001322968.1 NM_001322969.1 NM_001322970.1 NM_001322971.1 NM_001322974.1 XM_017016279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682923 c.607G>T p.G203* Chr.10: 124405477-124405477 on Build GRCh38
1603111 c.626A>G p.Y209C Chr.10: 124405458-124405458 on Build GRCh38
3699856 c.634C>T p.L212L Chr.10: 124405450-124405450 on Build GRCh38
6732103 c.583G>A p.D195N Chr.10: 124405501-124405501 on Build GRCh38
5929216 c.593G>A p.G198E Chr.10: 124405491-124405491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121965056 Chr.10: 124405457-124405457 on Build GRCh38
rs369018496 Chr.10: 124405569-124405569 on Build GRCh38
rs576876835 Chr.10: 124405570-124405570 on Build GRCh38
rs768951986 Chr.10: 124405589-124405606 on Build GRCh38
rs267606925 Chr.10: 124405488-124405488 on Build GRCh38
rs369806549 Chr.10: 124405478-124405478 on Build GRCh38
rs748602908 Chr.10: 124405518-124405518 on Build GRCh38
rs753377778 Chr.10: 124405499-124405499 on Build GRCh38
rs767272991 Chr.10: 124405586-124405586 on Build GRCh38
rs121965050 Chr.10: 124405534-124405534 on Build GRCh38
rs760896432 Chr.10: 124405446-124405446 on Build GRCh38
rs779470531 Chr.10: 124405423-124405423 on Build GRCh38
rs771151529 Chr.10: 124405539-124405539 on Build GRCh38
rs769824110 Chr.10: 124405555-124405555 on Build GRCh38
rs778189671 Chr.10: 124405531-124405531 on Build GRCh38
rs386833612 Chr.10: 124405548-124405552 on Build GRCh38
rs755162030 Chr.10: 124405451-124405451 on Build GRCh38
rs772528371 Chr.10: 124405424-124405424 on Build GRCh38
rs758705296 Chr.10: 124405529-124405529 on Build GRCh38
rs754489812 Chr.10: 124405502-124405502 on Build GRCh38
rs376636397 Chr.10: 124405575-124405578 on Build GRCh38
rs558611630 Chr.10: 124405541-124405541 on Build GRCh38
rs375157197 Chr.10: 124405470-124405470 on Build GRCh38
rs750055546 Chr.10: 124405472-124405472 on Build GRCh38
rs779597685 Chr.10: 124405506-124405506 on Build GRCh38
rs9422807 Chr.10: 124405422-124405422 on Build GRCh38
rs770977459 Chr.10: 124405454-124405454 on Build GRCh38
rs773314783 Chr.10: 124405445-124405445 on Build GRCh38
rs890106099 Chr.10: 124405550-124405550 on Build GRCh38
rs144763058 Chr.10: 124405538-124405538 on Build GRCh38
rs121965035 Chr.10: 124405532-124405534 on Build GRCh38
rs776703980 Chr.10: 124405479-124405479 on Build GRCh38
rs959408478 Chr.10: 124405437-124405437 on Build GRCh38
rs386833614 Chr.10: 124405501-124405501 on Build GRCh38
rs386833613 Chr.10: 124405542-124405542 on Build GRCh38
rs760295579 Chr.10: 124405596-124405596 on Build GRCh38
rs921694497 Chr.10: 124405515-124405515 on Build GRCh38
rs121965040 Chr.10: 124405545-124405545 on Build GRCh38
rs267606923 Chr.10: 124405551-124405551 on Build GRCh38
rs773030577 Chr.10: 124405591-124405591 on Build GRCh38
rs111616939 Chr.10: 124405441-124405441 on Build GRCh38
rs372492529 Chr.10: 124405471-124405471 on Build GRCh38
rs763993433 Chr.10: 124405461-124405461 on Build GRCh38
rs868567649 Chr.10: 124405480-124405480 on Build GRCh38
rs144823472 Chr.10: 124405469-124405469 on Build GRCh38

View Full Product Details