Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 118842941-118843170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EN1
Entrez Gene ID
2019
Gene Name
engrailed homeobox 1
Gene Aliases
-
UniGene
Hs.271977
Gene Chromosome Location
Chr.2: 118842171-118848183 on Build GRCh38
Transcripts
NM_001426.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3041073 c.1167C>T p.D389D Chr.2: 118842950-118842950 on Build GRCh38
4902725 c.1126G>A p.G376R Chr.2: 118842991-118842991 on Build GRCh38
3041076 c.1077G>A p.K359K Chr.2: 118843040-118843040 on Build GRCh38
6053427 c.1150A>T p.T384S Chr.2: 118842967-118842967 on Build GRCh38
4932863 c.1044G>T p.K348N Chr.2: 118843073-118843073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777982562 Chr.2: 118842982-118842982 on Build GRCh38
rs769343130 Chr.2: 118843025-118843025 on Build GRCh38
rs746164063 Chr.2: 118843017-118843017 on Build GRCh38
rs749585707 Chr.2: 118843000-118843000 on Build GRCh38
rs765343713 Chr.2: 118843024-118843024 on Build GRCh38
rs374991047 Chr.2: 118843106-118843106 on Build GRCh38
rs367986532 Chr.2: 118843112-118843112 on Build GRCh38
rs76483309 Chr.2: 118843088-118843088 on Build GRCh38
rs752734087 Chr.2: 118843071-118843071 on Build GRCh38
rs200272085 Chr.2: 118843054-118843054 on Build GRCh38
rs1042010097 Chr.2: 118843105-118843105 on Build GRCh38
rs201019085 Chr.2: 118843028-118843028 on Build GRCh38
rs752842693 Chr.2: 118842980-118842980 on Build GRCh38
rs751378198 Chr.2: 118843051-118843051 on Build GRCh38
rs1010055525 Chr.2: 118843096-118843096 on Build GRCh38
rs867601858 Chr.2: 118843108-118843108 on Build GRCh38
rs370237510 Chr.2: 118843055-118843055 on Build GRCh38
rs549340318 Chr.2: 118843109-118843109 on Build GRCh38
rs767392804 Chr.2: 118843058-118843058 on Build GRCh38
rs765877485 Chr.2: 118843049-118843049 on Build GRCh38
rs772694757 Chr.2: 118843027-118843027 on Build GRCh38
rs771057192 Chr.2: 118843001-118843001 on Build GRCh38
rs139806570 Chr.2: 118843019-118843019 on Build GRCh38
rs780372731 Chr.2: 118843148-118843148 on Build GRCh38
rs72955069 Chr.2: 118843009-118843009 on Build GRCh38
rs745951039 Chr.2: 118843023-118843023 on Build GRCh38
rs758801707 Chr.2: 118843130-118843130 on Build GRCh38
rs927334345 Chr.2: 118843045-118843045 on Build GRCh38
rs778069643 Chr.2: 118843089-118843089 on Build GRCh38
rs781213770 Chr.2: 118842962-118842962 on Build GRCh38

View Full Product Details