Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2396501-2396726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM6
Entrez Gene ID
343641
Gene Name
transglutaminase 6
Gene Aliases
SCA35, TG6, TGM3L, TGY, dJ734P14.3
UniGene
Hs.452039
Gene Chromosome Location
Chr.20: 2380908-2432753 on Build GRCh38
Transcripts
NM_001254734.1 NM_198994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7393695 c.513C>A p.H171Q Chr.20: 2396594-2396594 on Build GRCh38
6250943 c.442G>T p.A148S Chr.20: 2396523-2396523 on Build GRCh38
4518092 c.521_522CC>TT p.A174V Chr.20: 2396602-2396603 on Build GRCh38
6262139 c.430G>T p.D144Y Chr.20: 2396511-2396511 on Build GRCh38
1229115 c.478G>A p.D160N Chr.20: 2396559-2396559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771311792 Chr.20: 2396669-2396669 on Build GRCh38
rs559046657 Chr.20: 2396614-2396614 on Build GRCh38
rs868821371 Chr.20: 2396536-2396536 on Build GRCh38
rs779578465 Chr.20: 2396597-2396597 on Build GRCh38
rs748684872 Chr.20: 2396658-2396658 on Build GRCh38
rs780869334 Chr.20: 2396564-2396564 on Build GRCh38
rs543298933 Chr.20: 2396598-2396598 on Build GRCh38
rs748778105 Chr.20: 2396635-2396635 on Build GRCh38
rs1049973489 Chr.20: 2396665-2396665 on Build GRCh38
rs766655658 Chr.20: 2396603-2396603 on Build GRCh38
rs976949088 Chr.20: 2396696-2396696 on Build GRCh38
rs756760933 Chr.20: 2396623-2396624 on Build GRCh38
rs764045390 Chr.20: 2396561-2396561 on Build GRCh38
rs767673938 Chr.20: 2396523-2396523 on Build GRCh38
rs1017037115 Chr.20: 2396687-2396687 on Build GRCh38
rs758286096 Chr.20: 2396559-2396559 on Build GRCh38
rs45449695 Chr.20: 2396699-2396699 on Build GRCh38
rs373572401 Chr.20: 2396633-2396633 on Build GRCh38
rs755777005 Chr.20: 2396631-2396631 on Build GRCh38
rs772393406 Chr.20: 2396664-2396664 on Build GRCh38
rs201210375 Chr.20: 2396577-2396577 on Build GRCh38
rs772473600 Chr.20: 2396584-2396584 on Build GRCh38
rs779348597 Chr.20: 2396652-2396652 on Build GRCh38
rs866660553 Chr.20: 2396599-2396599 on Build GRCh38
rs16984872 Chr.20: 2396558-2396558 on Build GRCh38
rs967319494 Chr.20: 2396673-2396673 on Build GRCh38
rs776877060 Chr.20: 2396611-2396611 on Build GRCh38
rs755683440 Chr.20: 2396569-2396569 on Build GRCh38
rs140070681 Chr.20: 2396618-2396618 on Build GRCh38
rs778298713 Chr.20: 2396667-2396667 on Build GRCh38
rs147494925 Chr.20: 2396583-2396583 on Build GRCh38
rs776964940 Chr.20: 2396670-2396670 on Build GRCh38
rs757042943 Chr.20: 2396563-2396563 on Build GRCh38
rs932931072 Chr.20: 2396700-2396700 on Build GRCh38
rs759580224 Chr.20: 2396671-2396671 on Build GRCh38
rs371872705 Chr.20: 2396619-2396619 on Build GRCh38
rs774880823 Chr.20: 2396582-2396582 on Build GRCh38
rs781436768 Chr.20: 2396676-2396676 on Build GRCh38
rs141930579 Chr.20: 2396562-2396562 on Build GRCh38
rs767017450 Chr.20: 2396549-2396549 on Build GRCh38
rs750147778 Chr.20: 2396625-2396625 on Build GRCh38
rs760984105 Chr.20: 2396601-2396601 on Build GRCh38
rs767291936 Chr.20: 2396622-2396622 on Build GRCh38
rs779543685 Chr.20: 2396570-2396570 on Build GRCh38
rs992743590 Chr.20: 2396565-2396565 on Build GRCh38
rs878912863 Chr.20: 2396692-2396692 on Build GRCh38
rs766563849 Chr.20: 2396553-2396553 on Build GRCh38
rs150619818 Chr.20: 2396550-2396550 on Build GRCh38
rs370007894 Chr.20: 2396641-2396641 on Build GRCh38
rs755613950 Chr.20: 2396578-2396578 on Build GRCh38
rs375003299 Chr.20: 2396621-2396621 on Build GRCh38

View Full Product Details