Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3337944-3338217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSSC1
Entrez Gene ID
7260
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
UniGene
Hs.502770
Gene Chromosome Location
Chr.2: 3188968-3377882 on Build GRCh38
Transcripts
NM_003310.2 XM_011510383.2 XM_011510384.2 XM_011510385.2 XM_011510387.2 XM_017004818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810743 c.257G>T p.R86I Chr.2: 3338019-3338019 on Build GRCh38
6091253 c.252C>T p.Y84Y Chr.2: 3338024-3338024 on Build GRCh38
347630 c.203G>T p.W68L Chr.2: 3338073-3338073 on Build GRCh38
6091252 c.148G>T p.D50Y Chr.2: 3338128-3338128 on Build GRCh38
6009964 c.135C>A p.I45I Chr.2: 3338141-3338141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759289577 Chr.2: 3338041-3338041 on Build GRCh38
rs773102775 Chr.2: 3338086-3338086 on Build GRCh38
rs750762360 Chr.2: 3338159-3338159 on Build GRCh38
rs77169740 Chr.2: 3338139-3338139 on Build GRCh38
rs767325502 Chr.2: 3338116-3338118 on Build GRCh38
rs201909575 Chr.2: 3338189-3338189 on Build GRCh38
rs753059670 Chr.2: 3338062-3338062 on Build GRCh38
rs772272045 Chr.2: 3338070-3338070 on Build GRCh38
rs761598334 Chr.2: 3337986-3337986 on Build GRCh38
rs113205050 Chr.2: 3338174-3338174 on Build GRCh38
rs765670984 Chr.2: 3338157-3338157 on Build GRCh38
rs992329892 Chr.2: 3338094-3338094 on Build GRCh38
rs775584761 Chr.2: 3337994-3337994 on Build GRCh38
rs770698938 Chr.2: 3338096-3338096 on Build GRCh38
rs780172481 Chr.2: 3338077-3338077 on Build GRCh38
rs756532870 Chr.2: 3338191-3338202 on Build GRCh38
rs746172797 Chr.2: 3337975-3337975 on Build GRCh38
rs751167765 Chr.2: 3338034-3338034 on Build GRCh38
rs747272766 Chr.2: 3338168-3338168 on Build GRCh38
rs752311582 Chr.2: 3338121-3338121 on Build GRCh38
rs757784946 Chr.2: 3338147-3338147 on Build GRCh38
rs543868199 Chr.2: 3338033-3338033 on Build GRCh38
rs767089018 Chr.2: 3338042-3338042 on Build GRCh38
rs772035973 Chr.2: 3337981-3337981 on Build GRCh38
rs768773178 Chr.2: 3338085-3338085 on Build GRCh38
rs766899810 Chr.2: 3338111-3338111 on Build GRCh38
rs763732581 Chr.2: 3338136-3338136 on Build GRCh38
rs746235249 Chr.2: 3338066-3338066 on Build GRCh38
rs758713617 Chr.2: 3338160-3338160 on Build GRCh38
rs368506612 Chr.2: 3338183-3338186 on Build GRCh38
rs199764167 Chr.2: 3338044-3338044 on Build GRCh38
rs200591715 Chr.2: 3338009-3338009 on Build GRCh38
rs755046180 Chr.2: 3338172-3338172 on Build GRCh38
rs747064442 Chr.2: 3338084-3338084 on Build GRCh38
rs3211227 Chr.2: 3338055-3338055 on Build GRCh38
rs754149320 Chr.2: 3338183-3338183 on Build GRCh38
rs368182667 Chr.2: 3338047-3338047 on Build GRCh38
rs1011808434 Chr.2: 3338052-3338052 on Build GRCh38
rs199777526 Chr.2: 3338129-3338129 on Build GRCh38
rs148303243 Chr.2: 3338088-3338088 on Build GRCh38
rs371868745 Chr.2: 3338063-3338063 on Build GRCh38
rs778372222 Chr.2: 3338054-3338054 on Build GRCh38
rs754387878 Chr.2: 3338143-3338143 on Build GRCh38
rs773156071 Chr.2: 3338019-3338019 on Build GRCh38
rs779250972 Chr.2: 3337972-3337972 on Build GRCh38
rs200392582 Chr.2: 3338000-3338000 on Build GRCh38
rs375044108 Chr.2: 3338099-3338099 on Build GRCh38
rs916838083 Chr.2: 3338098-3338098 on Build GRCh38
rs149970040 Chr.2: 3338124-3338124 on Build GRCh38
rs148621025 Chr.2: 3338140-3338140 on Build GRCh38
rs780302609 Chr.2: 3338167-3338167 on Build GRCh38
rs370183545 Chr.2: 3338022-3338022 on Build GRCh38
rs561476823 Chr.2: 3338008-3338008 on Build GRCh38

View Full Product Details