Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20556504-20556755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASE9
Entrez Gene ID
390443
Gene Name
ribonuclease A family member 9 (inactive)
Gene Aliases
HEL128, RAK1, h461
UniGene
Hs.533972
Gene Chromosome Location
Chr.14: 20556093-20560931 on Build GRCh38
Transcripts
NM_001001673.3 NM_001110356.1 NM_001110357.1 NM_001110358.1 NM_001110359.1 NM_001110360.1 NM_001110361.1 NM_001289110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3361094 c.444T>C p.F148F Chr.14: 20556626-20556626 on Build GRCh38
5847767 c.562C>T p.P188S Chr.14: 20556508-20556508 on Build GRCh38
6054342 c.355A>T p.R119* Chr.14: 20556715-20556715 on Build GRCh38
3494711 c.356G>A p.R119K Chr.14: 20556714-20556714 on Build GRCh38
5614096 c.426T>A p.C142* Chr.14: 20556644-20556644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142708288 Chr.14: 20556599-20556599 on Build GRCh38
rs778344797 Chr.14: 20556595-20556595 on Build GRCh38
rs531497281 Chr.14: 20556702-20556702 on Build GRCh38
rs145447895 Chr.14: 20556569-20556569 on Build GRCh38
rs142287899 Chr.14: 20556647-20556647 on Build GRCh38
rs757877031 Chr.14: 20556666-20556666 on Build GRCh38
rs202187453 Chr.14: 20556630-20556630 on Build GRCh38
rs201519205 Chr.14: 20556708-20556708 on Build GRCh38
rs780634677 Chr.14: 20556542-20556542 on Build GRCh38
rs557776384 Chr.14: 20556693-20556693 on Build GRCh38
rs756093995 Chr.14: 20556586-20556586 on Build GRCh38
rs12590446 Chr.14: 20556627-20556627 on Build GRCh38
rs111600594 Chr.14: 20556605-20556605 on Build GRCh38
rs767725432 Chr.14: 20556578-20556578 on Build GRCh38
rs772956589 Chr.14: 20556534-20556534 on Build GRCh38
rs759335294 Chr.14: 20556723-20556723 on Build GRCh38
rs950228882 Chr.14: 20556561-20556561 on Build GRCh38
rs560127702 Chr.14: 20556614-20556614 on Build GRCh38
rs766275623 Chr.14: 20556626-20556626 on Build GRCh38
rs750875521 Chr.14: 20556676-20556676 on Build GRCh38
rs761224794 Chr.14: 20556529-20556529 on Build GRCh38
rs139929154 Chr.14: 20556541-20556541 on Build GRCh38
rs939318974 Chr.14: 20556619-20556619 on Build GRCh38
rs141963632 Chr.14: 20556613-20556613 on Build GRCh38
rs78916107 Chr.14: 20556703-20556703 on Build GRCh38
rs373837965 Chr.14: 20556581-20556581 on Build GRCh38
rs752791748 Chr.14: 20556652-20556652 on Build GRCh38
rs147407456 Chr.14: 20556540-20556540 on Build GRCh38
rs765277588 Chr.14: 20556532-20556532 on Build GRCh38
rs780929947 Chr.14: 20556689-20556689 on Build GRCh38
rs758777788 Chr.14: 20556686-20556686 on Build GRCh38
rs779556749 Chr.14: 20556667-20556667 on Build GRCh38
rs893488734 Chr.14: 20556611-20556611 on Build GRCh38
rs892526272 Chr.14: 20556635-20556635 on Build GRCh38
rs574617651 Chr.14: 20556609-20556609 on Build GRCh38
rs762928729 Chr.14: 20556621-20556621 on Build GRCh38
rs770915786 Chr.14: 20556714-20556714 on Build GRCh38
rs753929887 Chr.14: 20556661-20556661 on Build GRCh38
rs151318768 Chr.14: 20556658-20556658 on Build GRCh38
rs554647850 Chr.14: 20556616-20556616 on Build GRCh38
rs779466707 Chr.14: 20556604-20556604 on Build GRCh38
rs776319948 Chr.14: 20556528-20556528 on Build GRCh38
rs755236091 Chr.14: 20556577-20556577 on Build GRCh38
rs529014375 Chr.14: 20556716-20556716 on Build GRCh38
rs748283218 Chr.14: 20556580-20556580 on Build GRCh38

View Full Product Details