Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55635497-55635722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A9
Entrez Gene ID
153129
Gene Name
solute carrier family 38 member 9
Gene Aliases
URLC11
UniGene
Hs.742101
Gene Chromosome Location
Chr.5: 55625845-55712344 on Build GRCh38
Transcripts
NM_001258286.1 NM_001258287.1 NM_001282429.1 NM_173514.3 NR_047649.1 XM_006714537.2 XM_006714538.3 XM_006714539.3 XM_011543173.1 XM_011543174.1 XM_011543176.1 XM_011543177.1 XM_011543178.1 XM_011543180.1 XM_011543182.1 XM_011543183.1 XM_011543184.1 XM_017009077.1 XM_017009078.1 XM_017009079.1 XM_017009080.1 XM_017009081.1 XM_017009082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741336 c.1276G>C p.E426Q Chr.5: 55635549-55635549 on Build GRCh38
4778120 c.1173G>T p.R391S Chr.5: 55635652-55635652 on Build GRCh38
4778119 c.1173G>T p.R391S Chr.5: 55635652-55635652 on Build GRCh38
1620398 c.1259_1260insT p.L420fs*7 Chr.5: 55635565-55635566 on Build GRCh38
449776 c.1170G>A p.V390V Chr.5: 55635655-55635655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922529741 Chr.5: 55635689-55635689 on Build GRCh38
rs571739753 Chr.5: 55635678-55635678 on Build GRCh38
rs925686178 Chr.5: 55635633-55635633 on Build GRCh38
rs775950762 Chr.5: 55635607-55635607 on Build GRCh38
rs752385378 Chr.5: 55635529-55635529 on Build GRCh38
rs758245774 Chr.5: 55635533-55635533 on Build GRCh38
rs752721116 Chr.5: 55635626-55635626 on Build GRCh38
rs770151453 Chr.5: 55635549-55635549 on Build GRCh38
rs778112362 Chr.5: 55635534-55635534 on Build GRCh38
rs370969665 Chr.5: 55635627-55635627 on Build GRCh38
rs760389921 Chr.5: 55635606-55635606 on Build GRCh38
rs772835868 Chr.5: 55635586-55635586 on Build GRCh38
rs373829901 Chr.5: 55635682-55635682 on Build GRCh38
rs757406359 Chr.5: 55635539-55635539 on Build GRCh38
rs764045623 Chr.5: 55635639-55635639 on Build GRCh38
rs780637971 Chr.5: 55635559-55635559 on Build GRCh38
rs754475264 Chr.5: 55635594-55635594 on Build GRCh38
rs142135949 Chr.5: 55635645-55635645 on Build GRCh38
rs781352452 Chr.5: 55635540-55635540 on Build GRCh38
rs779114329 Chr.5: 55635629-55635636 on Build GRCh38
rs767802162 Chr.5: 55635681-55635681 on Build GRCh38
rs745972355 Chr.5: 55635545-55635545 on Build GRCh38
rs769055013 Chr.5: 55635579-55635579 on Build GRCh38
rs759112987 Chr.5: 55635611-55635611 on Build GRCh38
rs148631803 Chr.5: 55635570-55635570 on Build GRCh38
rs751881735 Chr.5: 55635538-55635538 on Build GRCh38
rs765046146 Chr.5: 55635617-55635617 on Build GRCh38

View Full Product Details