Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4055159-4055406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB7A
Entrez Gene ID
51341
Gene Name
zinc finger and BTB domain containing 7A
Gene Aliases
FBI-1, FBI1, LRF, TIP21, ZBTB7, ZNF857A, pokemon
UniGene
Hs.591384
Gene Chromosome Location
Chr.19: 4043304-4066945 on Build GRCh38
Transcripts
NM_001317990.1 NM_015898.3 XM_005259570.4 XM_005259571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6580997 c.36G>A p.P12P Chr.19: 4055197-4055197 on Build GRCh38
5986713 c.73_74AA>TC p.N25S Chr.19: 4055159-4055160 on Build GRCh38
6825459 c.51C>A p.S17R Chr.19: 4055182-4055182 on Build GRCh38
3533986 c.40C>T p.P14S Chr.19: 4055193-4055193 on Build GRCh38
4492565 c.39C>T p.F13F Chr.19: 4055194-4055194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891764726 Chr.19: 4055343-4055343 on Build GRCh38
rs770385730 Chr.19: 4055179-4055179 on Build GRCh38
rs760323797 Chr.19: 4055262-4055262 on Build GRCh38
rs1043564657 Chr.19: 4055306-4055306 on Build GRCh38
rs774351119 Chr.19: 4055211-4055211 on Build GRCh38
rs538559850 Chr.19: 4055283-4055283 on Build GRCh38
rs146580794 Chr.19: 4055276-4055276 on Build GRCh38
rs575381525 Chr.19: 4055330-4055330 on Build GRCh38
rs546189028 Chr.19: 4055265-4055265 on Build GRCh38
rs1026683298 Chr.19: 4055310-4055310 on Build GRCh38
rs1012931799 Chr.19: 4055215-4055215 on Build GRCh38
rs750331701 Chr.19: 4055243-4055243 on Build GRCh38
rs1003505874 Chr.19: 4055227-4055227 on Build GRCh38
rs556775501 Chr.19: 4055277-4055277 on Build GRCh38
rs1053135099 Chr.19: 4055339-4055339 on Build GRCh38
rs1015789906 Chr.19: 4055239-4055239 on Build GRCh38
rs953761771 Chr.19: 4055320-4055320 on Build GRCh38
rs866053656 Chr.19: 4055228-4055228 on Build GRCh38
rs749763962 Chr.19: 4055376-4055376 on Build GRCh38
rs1054544052 Chr.19: 4055200-4055200 on Build GRCh38
rs200936052 Chr.19: 4055224-4055224 on Build GRCh38
rs757069218 Chr.19: 4055226-4055226 on Build GRCh38
rs766474113 Chr.19: 4055209-4055209 on Build GRCh38
rs752165308 Chr.19: 4055387-4055387 on Build GRCh38
rs763595294 Chr.19: 4055206-4055206 on Build GRCh38
rs536240873 Chr.19: 4055245-4055245 on Build GRCh38
rs755657227 Chr.19: 4055388-4055388 on Build GRCh38
rs149842256 Chr.19: 4055197-4055197 on Build GRCh38
rs550633705 Chr.19: 4055221-4055221 on Build GRCh38
rs117368092 Chr.19: 4055327-4055327 on Build GRCh38
rs765070162 Chr.19: 4055240-4055240 on Build GRCh38

View Full Product Details