Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 107602292-107602561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV3
Entrez Gene ID
10451
Gene Name
vav guanine nucleotide exchange factor 3
Gene Aliases
-
UniGene
Hs.267659
Gene Chromosome Location
Chr.1: 107571160-107964923 on Build GRCh38
Transcripts
NM_001079874.1 NM_006113.4 XM_005270360.2 XM_017000053.1 XM_017000054.1 XM_017000055.1 XM_017000056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4620902 c.2220G>C p.M740I Chr.1: 107602397-107602397 on Build GRCh38
6819594 c.524A>T p.K175I Chr.1: 107602413-107602413 on Build GRCh38
6819593 c.2204A>T p.K735I Chr.1: 107602413-107602413 on Build GRCh38
2116628 c.2151G>A p.K717K Chr.1: 107602466-107602466 on Build GRCh38
5792327 c.2194G>C p.E732Q Chr.1: 107602423-107602423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575375101 Chr.1: 107602393-107602393 on Build GRCh38
rs374741440 Chr.1: 107602506-107602506 on Build GRCh38
rs776429384 Chr.1: 107602418-107602418 on Build GRCh38
rs765425486 Chr.1: 107602499-107602499 on Build GRCh38
rs754077861 Chr.1: 107602462-107602462 on Build GRCh38
rs757055871 Chr.1: 107602365-107602365 on Build GRCh38
rs943261463 Chr.1: 107602492-107602492 on Build GRCh38
rs751354243 Chr.1: 107602362-107602362 on Build GRCh38
rs558440664 Chr.1: 107602340-107602340 on Build GRCh38
rs749444215 Chr.1: 107602372-107602372 on Build GRCh38
rs755336967 Chr.1: 107602461-107602461 on Build GRCh38
rs370224254 Chr.1: 107602518-107602518 on Build GRCh38
rs1040701747 Chr.1: 107602501-107602501 on Build GRCh38
rs762497588 Chr.1: 107602444-107602444 on Build GRCh38
rs771321097 Chr.1: 107602415-107602415 on Build GRCh38
rs747481089 Chr.1: 107602520-107602520 on Build GRCh38
rs544452793 Chr.1: 107602460-107602460 on Build GRCh38
rs780201846 Chr.1: 107602367-107602367 on Build GRCh38
rs949866970 Chr.1: 107602517-107602517 on Build GRCh38
rs761002099 Chr.1: 107602411-107602411 on Build GRCh38
rs777087709 Chr.1: 107602532-107602532 on Build GRCh38
rs768733946 Chr.1: 107602373-107602373 on Build GRCh38
rs916985316 Chr.1: 107602512-107602512 on Build GRCh38
rs1041804026 Chr.1: 107602531-107602531 on Build GRCh38
rs773746469 Chr.1: 107602403-107602403 on Build GRCh38
rs971838859 Chr.1: 107602368-107602368 on Build GRCh38
rs778912131 Chr.1: 107602391-107602391 on Build GRCh38
rs764863269 Chr.1: 107602429-107602429 on Build GRCh38
rs757068688 Chr.1: 107602457-107602457 on Build GRCh38
rs951641047 Chr.1: 107602490-107602490 on Build GRCh38
rs752215005 Chr.1: 107602442-107602442 on Build GRCh38
rs759208923 Chr.1: 107602426-107602426 on Build GRCh38
rs371750799 Chr.1: 107602478-107602478 on Build GRCh38
rs755149974 Chr.1: 107602463-107602463 on Build GRCh38
rs772177850 Chr.1: 107602510-107602510 on Build GRCh38
rs763990917 Chr.1: 107602449-107602449 on Build GRCh38
rs772495306 Chr.1: 107602394-107602394 on Build GRCh38
rs751504688 Chr.1: 107602456-107602456 on Build GRCh38
rs1018259093 Chr.1: 107602326-107602326 on Build GRCh38

View Full Product Details