Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 607328-607553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101926963
Entrez Gene ID
101926963
Gene Name
uncharacterized LOC101926963
Gene Aliases
-
UniGene
Hs.521338
Gene Chromosome Location
Chr.7: 602845-608502 on Build GRCh38
Transcripts
NR_110054.1 NR_110055.1 NM_001164758.1 NM_001164759.1 NM_001164760.1 NM_001164761.1 NM_001164762.1 NM_002735.2 XM_011515446.2 XM_017012434.1

Gene Symbol
PRKAR1B
Entrez Gene ID
5575
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
Gene Aliases
PRKAR1
UniGene
Hs.520851
Gene Chromosome Location
Chr.7: 549185-727676 on Build GRCh38
Transcripts
NR_110054.1 NR_110055.1 NM_001164758.1 NM_001164759.1 NM_001164760.1 NM_001164761.1 NM_001164762.1 NM_002735.2 XM_011515446.2 XM_017012434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361774 c.491T>A p.V164D Chr.7: 607402-607402 on Build GRCh38
4717677 c.478G>A p.A160T Chr.7: 607415-607415 on Build GRCh38
4717676 c.493A>G p.I165V Chr.7: 607400-607400 on Build GRCh38
1263052 c.450C>T p.F150F Chr.7: 607443-607443 on Build GRCh38
3085454 c.462C>T p.F154F Chr.7: 607431-607431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773154465 Chr.7: 607416-607416 on Build GRCh38
rs752375402 Chr.7: 607369-607369 on Build GRCh38
rs371840421 Chr.7: 607493-607493 on Build GRCh38
rs748546239 Chr.7: 607406-607406 on Build GRCh38
rs371872376 Chr.7: 607425-607425 on Build GRCh38
rs561731404 Chr.7: 607370-607370 on Build GRCh38
rs758056313 Chr.7: 607466-607466 on Build GRCh38
rs749324714 Chr.7: 607495-607495 on Build GRCh38
rs781597164 Chr.7: 607485-607485 on Build GRCh38
rs764777147 Chr.7: 607443-607443 on Build GRCh38
rs781693548 Chr.7: 607385-607385 on Build GRCh38
rs374524811 Chr.7: 607371-607371 on Build GRCh38
rs564291448 Chr.7: 607521-607521 on Build GRCh38
rs910955134 Chr.7: 607498-607498 on Build GRCh38
rs530747068 Chr.7: 607387-607387 on Build GRCh38
rs776586095 Chr.7: 607430-607430 on Build GRCh38
rs141011841 Chr.7: 607404-607404 on Build GRCh38
rs973721293 Chr.7: 607468-607468 on Build GRCh38
rs199730179 Chr.7: 607364-607364 on Build GRCh38
rs777352655 Chr.7: 607490-607490 on Build GRCh38
rs745954573 Chr.7: 607386-607386 on Build GRCh38
rs980352358 Chr.7: 607376-607376 on Build GRCh38
rs376223777 Chr.7: 607461-607461 on Build GRCh38
rs367970366 Chr.7: 607479-607479 on Build GRCh38
rs746352943 Chr.7: 607486-607486 on Build GRCh38
rs754472631 Chr.7: 607355-607355 on Build GRCh38
rs747313329 Chr.7: 607373-607373 on Build GRCh38
rs775613517 Chr.7: 607389-607389 on Build GRCh38
rs1006169667 Chr.7: 607487-607487 on Build GRCh38
rs763666358 Chr.7: 607469-607469 on Build GRCh38
rs543771352 Chr.7: 607518-607518 on Build GRCh38
rs771280762 Chr.7: 607456-607456 on Build GRCh38
rs368437414 Chr.7: 607377-607377 on Build GRCh38
rs759019649 Chr.7: 607442-607442 on Build GRCh38
rs772049509 Chr.7: 607415-607415 on Build GRCh38

View Full Product Details