Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51941016-51941289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP7B
Entrez Gene ID
540
Gene Name
ATPase copper transporting beta
Gene Aliases
PWD, WC1, WD, WND
UniGene
Hs.492280
Gene Chromosome Location
Chr.13: 51932669-52012130 on Build GRCh38
Transcripts
NM_000053.3 NM_001005918.2 NM_001243182.1 XM_005266423.2 XM_005266424.4 XM_005266427.2 XM_005266428.1 XM_005266430.4 XM_005266431.3 XM_005266432.2 XM_006719837.3 XM_006719838.1 XM_006719839.1 XM_011535117.2 XM_011535118.1 XM_011535119.1 XM_011535122.2 XM_017020627.1 XM_017020628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1562211 c.3532A>G p.T1178A Chr.13: 51941105-51941105 on Build GRCh38
3748575 c.3466C>T p.R1156C Chr.13: 51941171-51941171 on Build GRCh38
4048035 c.3452G>A p.R1151H Chr.13: 51941185-51941185 on Build GRCh38
3469254 c.3450C>T p.N1150N Chr.13: 51941187-51941187 on Build GRCh38
4048034 c.3502G>A p.A1168T Chr.13: 51941135-51941135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748924063 Chr.13: 51941084-51941085 on Build GRCh38
rs60431989 Chr.13: 51941194-51941194 on Build GRCh38
rs760199892 Chr.13: 51941238-51941238 on Build GRCh38
rs377297166 Chr.13: 51941185-51941185 on Build GRCh38
rs767069278 Chr.13: 51941175-51941175 on Build GRCh38
rs777600732 Chr.13: 51941080-51941081 on Build GRCh38
rs754931215 Chr.13: 51941246-51941246 on Build GRCh38
rs587783313 Chr.13: 51941209-51941209 on Build GRCh38
rs752712932 Chr.13: 51941070-51941070 on Build GRCh38
rs763963568 Chr.13: 51941136-51941136 on Build GRCh38
rs753108142 Chr.13: 51941065-51941065 on Build GRCh38
rs781499175 Chr.13: 51941217-51941217 on Build GRCh38
rs754759064 Chr.13: 51941203-51941203 on Build GRCh38
rs137853279 Chr.13: 51941111-51941111 on Build GRCh38
rs765139243 Chr.13: 51941089-51941090 on Build GRCh38
rs750407028 Chr.13: 51941117-51941117 on Build GRCh38
rs765833446 Chr.13: 51941240-51941240 on Build GRCh38
rs757549770 Chr.13: 51941215-51941215 on Build GRCh38
rs908231616 Chr.13: 51941094-51941094 on Build GRCh38
rs765755993 Chr.13: 51941061-51941061 on Build GRCh38
rs776086518 Chr.13: 51941165-51941165 on Build GRCh38
rs372318135 Chr.13: 51941158-51941158 on Build GRCh38
rs764481833 Chr.13: 51941166-51941166 on Build GRCh38
rs751819044 Chr.13: 51941237-51941237 on Build GRCh38
rs201985951 Chr.13: 51941172-51941172 on Build GRCh38
rs893143787 Chr.13: 51941174-51941174 on Build GRCh38
rs587783314 Chr.13: 51941139-51941139 on Build GRCh38
rs753759128 Chr.13: 51941197-51941197 on Build GRCh38
rs756029120 Chr.13: 51941120-51941120 on Build GRCh38
rs768438514 Chr.13: 51941133-51941133 on Build GRCh38
rs867107727 Chr.13: 51941147-51941147 on Build GRCh38
rs765009591 Chr.13: 51941256-51941256 on Build GRCh38
rs767328462 Chr.13: 51941042-51941042 on Build GRCh38
rs369312119 Chr.13: 51941235-51941235 on Build GRCh38
rs786204547 Chr.13: 51941081-51941081 on Build GRCh38
rs760098934 Chr.13: 51941059-51941059 on Build GRCh38
rs761435604 Chr.13: 51941230-51941230 on Build GRCh38
rs749085322 Chr.13: 51941132-51941132 on Build GRCh38
rs940997440 Chr.13: 51941113-51941113 on Build GRCh38
rs780648361 Chr.13: 51941096-51941096 on Build GRCh38
rs193922106 Chr.13: 51941148-51941148 on Build GRCh38
rs1057516479 Chr.13: 51941108-51941108 on Build GRCh38
rs528866491 Chr.13: 51941121-51941121 on Build GRCh38
rs778749563 Chr.13: 51941211-51941211 on Build GRCh38
rs756751716 Chr.13: 51941082-51941082 on Build GRCh38
rs763671539 Chr.13: 51941078-51941078 on Build GRCh38
rs867501285 Chr.13: 51941212-51941213 on Build GRCh38
rs199924281 Chr.13: 51941221-51941221 on Build GRCh38
rs773415651 Chr.13: 51941051-51941051 on Build GRCh38
rs375226851 Chr.13: 51941055-51941055 on Build GRCh38
rs531370978 Chr.13: 51941259-51941259 on Build GRCh38
rs995516204 Chr.13: 51941114-51941114 on Build GRCh38
rs770428835 Chr.13: 51941164-51941164 on Build GRCh38
rs755817220 Chr.13: 51941086-51941086 on Build GRCh38
rs774844173 Chr.13: 51941230-51941231 on Build GRCh38
rs865803089 Chr.13: 51941144-51941144 on Build GRCh38
rs777320373 Chr.13: 51941058-51941058 on Build GRCh38
rs760682015 Chr.13: 51941054-51941054 on Build GRCh38
rs575346789 Chr.13: 51941047-51941048 on Build GRCh38
rs1057519121 Chr.13: 51941087-51941087 on Build GRCh38
rs755554442 Chr.13: 51941186-51941186 on Build GRCh38
rs371517600 Chr.13: 51941068-51941068 on Build GRCh38
rs184388696 Chr.13: 51941080-51941080 on Build GRCh38
rs763314617 Chr.13: 51941076-51941076 on Build GRCh38
rs1801249 Chr.13: 51941218-51941218 on Build GRCh38
rs587783315 Chr.13: 51941089-51941089 on Build GRCh38
rs773917820 Chr.13: 51941170-51941170 on Build GRCh38
rs375721306 Chr.13: 51941224-51941224 on Build GRCh38
rs753758073 Chr.13: 51941244-51941244 on Build GRCh38
rs769949202 Chr.13: 51941220-51941220 on Build GRCh38
rs747255077 Chr.13: 51941213-51941213 on Build GRCh38
rs771202253 Chr.13: 51941128-51941128 on Build GRCh38
rs373455989 Chr.13: 51941257-51941257 on Build GRCh38

View Full Product Details