Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87085707-87085948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGA
Entrez Gene ID
1081
Gene Name
glycoprotein hormones, alpha polypeptide
Gene Aliases
CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA
UniGene
Hs.119689
Gene Chromosome Location
Chr.6: 87085498-87095147 on Build GRCh38
Transcripts
NM_000735.3 NM_001252383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200910 c.275T>C p.V92A Chr.6: 87085832-87085832 on Build GRCh38
6107608 c.340C>A p.H114N Chr.6: 87085767-87085767 on Build GRCh38
70161 c.287G>C p.G96A Chr.6: 87085820-87085820 on Build GRCh38
4836802 c.340C>G p.H114D Chr.6: 87085767-87085767 on Build GRCh38
743971 c.338A>T p.Y113F Chr.6: 87085769-87085769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368530753 Chr.6: 87085856-87085856 on Build GRCh38
rs746918390 Chr.6: 87085847-87085847 on Build GRCh38
rs748376783 Chr.6: 87085865-87085865 on Build GRCh38
rs781214526 Chr.6: 87085863-87085863 on Build GRCh38
rs746839263 Chr.6: 87085908-87085908 on Build GRCh38
rs771897260 Chr.6: 87085880-87085880 on Build GRCh38
rs756147637 Chr.6: 87085867-87085867 on Build GRCh38
rs143780652 Chr.6: 87085839-87085839 on Build GRCh38
rs139810378 Chr.6: 87085828-87085828 on Build GRCh38
rs769877495 Chr.6: 87085770-87085770 on Build GRCh38
rs777807459 Chr.6: 87085872-87085872 on Build GRCh38
rs145923049 Chr.6: 87085795-87085795 on Build GRCh38
rs202123656 Chr.6: 87085823-87085823 on Build GRCh38
rs992172113 Chr.6: 87085761-87085761 on Build GRCh38
rs757113041 Chr.6: 87085856-87085856 on Build GRCh38
rs777802853 Chr.6: 87085846-87085847 on Build GRCh38
rs887588867 Chr.6: 87085898-87085898 on Build GRCh38
rs775554062 Chr.6: 87085883-87085883 on Build GRCh38
rs760584468 Chr.6: 87085820-87085820 on Build GRCh38
rs977795099 Chr.6: 87085836-87085836 on Build GRCh38
rs748215278 Chr.6: 87085755-87085755 on Build GRCh38
rs373897638 Chr.6: 87085735-87085735 on Build GRCh38
rs201187778 Chr.6: 87085779-87085779 on Build GRCh38
rs768732919 Chr.6: 87085737-87085737 on Build GRCh38
rs142673245 Chr.6: 87085792-87085792 on Build GRCh38
rs749409684 Chr.6: 87085878-87085878 on Build GRCh38
rs781412283 Chr.6: 87085876-87085877 on Build GRCh38
rs758439462 Chr.6: 87085747-87085748 on Build GRCh38
rs918005050 Chr.6: 87085771-87085771 on Build GRCh38
rs77122732 Chr.6: 87085850-87085850 on Build GRCh38
rs372445287 Chr.6: 87085873-87085876 on Build GRCh38
rs781547088 Chr.6: 87085751-87085751 on Build GRCh38
rs951460114 Chr.6: 87085740-87085740 on Build GRCh38
rs763639075 Chr.6: 87085822-87085822 on Build GRCh38
rs372856498 Chr.6: 87085887-87085887 on Build GRCh38
rs888880439 Chr.6: 87085911-87085911 on Build GRCh38
rs758954772 Chr.6: 87085847-87085847 on Build GRCh38
rs745838602 Chr.6: 87085793-87085793 on Build GRCh38
rs750908292 Chr.6: 87085845-87085845 on Build GRCh38
rs573675107 Chr.6: 87085739-87085739 on Build GRCh38
rs775116542 Chr.6: 87085818-87085818 on Build GRCh38
rs980809832 Chr.6: 87085827-87085827 on Build GRCh38
rs972292704 Chr.6: 87085731-87085731 on Build GRCh38
rs565869558 Chr.6: 87085872-87085873 on Build GRCh38

View Full Product Details