Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37625738-37625983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX52
Entrez Gene ID
11056
Gene Name
DEAD-box helicase 52
Gene Aliases
HUSSY19, ROK1
UniGene
Hs.590937
Gene Chromosome Location
Chr.17: 37609739-37643464 on Build GRCh38
Transcripts
NM_001291476.1 NM_007010.4 XM_011524232.2 XM_011524233.2

SNP Detail

SNP ID Chromosome Location
rs61749870 Chr.17: 37625937-37625937 on Build GRCh38
rs754387825 Chr.17: 37625887-37625887 on Build GRCh38
rs768440587 Chr.17: 37625940-37625940 on Build GRCh38
rs765607708 Chr.17: 37625896-37625896 on Build GRCh38
rs568352709 Chr.17: 37625794-37625794 on Build GRCh38
rs922233573 Chr.17: 37625925-37625925 on Build GRCh38
rs376605490 Chr.17: 37625939-37625939 on Build GRCh38
rs890839032 Chr.17: 37625874-37625874 on Build GRCh38
rs777910042 Chr.17: 37625917-37625917 on Build GRCh38
rs758810819 Chr.17: 37625903-37625903 on Build GRCh38
rs1012252953 Chr.17: 37625808-37625808 on Build GRCh38
rs770126415 Chr.17: 37625818-37625818 on Build GRCh38
rs755489849 Chr.17: 37625888-37625888 on Build GRCh38
rs767030051 Chr.17: 37625847-37625847 on Build GRCh38
rs767385481 Chr.17: 37625852-37625852 on Build GRCh38
rs140006660 Chr.17: 37625859-37625859 on Build GRCh38
rs1003390468 Chr.17: 37625846-37625846 on Build GRCh38
rs137989328 Chr.17: 37625922-37625922 on Build GRCh38
rs534347171 Chr.17: 37625797-37625797 on Build GRCh38
rs747841191 Chr.17: 37625955-37625955 on Build GRCh38
rs547576759 Chr.17: 37625838-37625838 on Build GRCh38
rs200253582 Chr.17: 37625883-37625883 on Build GRCh38
rs752961463 Chr.17: 37625898-37625898 on Build GRCh38
rs201448621 Chr.17: 37625851-37625852 on Build GRCh38
rs774370402 Chr.17: 37625947-37625947 on Build GRCh38
rs887061938 Chr.17: 37625806-37625806 on Build GRCh38
rs773013105 Chr.17: 37625852-37625852 on Build GRCh38
rs147610462 Chr.17: 37625851-37625851 on Build GRCh38

View Full Product Details