Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10774627-10774890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C2
Entrez Gene ID
245973
Gene Name
ATPase H+ transporting V1 subunit C2
Gene Aliases
ATP6C2, VMA5
UniGene
Hs.580464
Gene Chromosome Location
Chr.2: 10720974-10785110 on Build GRCh38
Transcripts
NM_001039362.1 NM_144583.3 XM_011510339.2 XM_011510340.2 XM_011510341.2 XM_017003745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4840884 c.724G>T p.E242* Chr.2: 10774873-10774873 on Build GRCh38
1184127 c.689G>A p.R230Q Chr.2: 10774838-10774838 on Build GRCh38
5214959 c.688C>T p.R230* Chr.2: 10774837-10774837 on Build GRCh38
3783326 c.672C>A p.F224L Chr.2: 10774821-10774821 on Build GRCh38
4840883 c.724G>T p.E242* Chr.2: 10774873-10774873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937626501 Chr.2: 10774678-10774678 on Build GRCh38
rs544695075 Chr.2: 10774842-10774842 on Build GRCh38
rs773670939 Chr.2: 10774844-10774844 on Build GRCh38
rs895995069 Chr.2: 10774716-10774716 on Build GRCh38
rs191315500 Chr.2: 10774796-10774796 on Build GRCh38
rs927260496 Chr.2: 10774648-10774648 on Build GRCh38
rs749243290 Chr.2: 10774771-10774771 on Build GRCh38
rs775004291 Chr.2: 10774794-10774794 on Build GRCh38
rs755102383 Chr.2: 10774739-10774739 on Build GRCh38
rs778792446 Chr.2: 10774783-10774783 on Build GRCh38
rs527493964 Chr.2: 10774837-10774837 on Build GRCh38
rs554946815 Chr.2: 10774700-10774700 on Build GRCh38
rs768171372 Chr.2: 10774814-10774814 on Build GRCh38
rs146332957 Chr.2: 10774849-10774849 on Build GRCh38
rs1057374808 Chr.2: 10774699-10774699 on Build GRCh38
rs1042257120 Chr.2: 10774725-10774725 on Build GRCh38
rs773215312 Chr.2: 10774718-10774718 on Build GRCh38
rs757222626 Chr.2: 10774775-10774775 on Build GRCh38
rs751699539 Chr.2: 10774738-10774738 on Build GRCh38
rs573150614 Chr.2: 10774703-10774703 on Build GRCh38
rs763055354 Chr.2: 10774729-10774729 on Build GRCh38
rs771801596 Chr.2: 10774793-10774793 on Build GRCh38
rs922962915 Chr.2: 10774709-10774709 on Build GRCh38
rs201869722 Chr.2: 10774815-10774815 on Build GRCh38
rs946250320 Chr.2: 10774717-10774717 on Build GRCh38
rs746491327 Chr.2: 10774829-10774830 on Build GRCh38
rs745628657 Chr.2: 10774792-10774792 on Build GRCh38
rs56195743 Chr.2: 10774741-10774741 on Build GRCh38

View Full Product Details