Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 63655393-63655620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF9
Entrez Gene ID
23229
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
UniGene
Hs.54697
Gene Chromosome Location
Chr.X: 63634967-63786025 on Build GRCh38
Transcripts
NM_001173479.1 NM_001173480.1 NM_015185.2 XM_005262249.2 XM_005262250.3 XM_005262251.1 XM_005262252.1 XM_011530890.1 XM_011530892.1 XM_017029363.1 XM_017029364.1 XM_017029365.1 XM_017029366.1 XM_017029367.1 XM_017029368.1 XM_017029369.1 XM_017029370.1 XM_017029371.1 XM_017029372.1 XM_017029373.1 XM_017029374.1 XM_017029375.1 XM_017029377.1 XM_017029378.1 XM_017029379.1 XM_017029380.1

SNP Detail

SNP ID Chromosome Location
rs782619006 Chr.X: 63655450-63655450 on Build GRCh38
rs781945520 Chr.X: 63655549-63655549 on Build GRCh38
rs782387990 Chr.X: 63655522-63655522 on Build GRCh38
rs56110425 Chr.X: 63655590-63655590 on Build GRCh38
rs782176476 Chr.X: 63655462-63655462 on Build GRCh38
rs782648725 Chr.X: 63655478-63655478 on Build GRCh38
rs782304484 Chr.X: 63655563-63655563 on Build GRCh38
rs781824011 Chr.X: 63655448-63655448 on Build GRCh38
rs782707712 Chr.X: 63655421-63655421 on Build GRCh38
rs782150415 Chr.X: 63655579-63655579 on Build GRCh38
rs782486555 Chr.X: 63655463-63655463 on Build GRCh38
rs782224124 Chr.X: 63655551-63655551 on Build GRCh38
rs370096217 Chr.X: 63655468-63655468 on Build GRCh38
rs782643207 Chr.X: 63655510-63655510 on Build GRCh38
rs781947373 Chr.X: 63655570-63655570 on Build GRCh38

View Full Product Details