Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89009155-89009428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP3
Entrez Gene ID
2635
Gene Name
guanylate binding protein 3
Gene Aliases
-
UniGene
Hs.720167
Gene Chromosome Location
Chr.1: 89006677-89022880 on Build GRCh38
Transcripts
NM_001319179.1 NM_001319180.1 NM_001319181.1 NM_018284.2 NR_135007.1 XM_005270743.2 XM_006710574.2 XM_011541233.1 XM_011541234.1 XM_017001013.1 XM_017001014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172175 c.1447A>C p.K483Q Chr.1: 89009410-89009410 on Build GRCh38
3706079 c.1459A>G p.I487V Chr.1: 89009398-89009398 on Build GRCh38
4596438 c.1445A>G p.E482G Chr.1: 89009412-89009412 on Build GRCh38
4980606 c.1432C>G p.Q478E Chr.1: 89009425-89009425 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769879227 Chr.1: 89009355-89009356 on Build GRCh38
rs576596322 Chr.1: 89009227-89009227 on Build GRCh38
rs776657841 Chr.1: 89009184-89009184 on Build GRCh38
rs981778901 Chr.1: 89009341-89009341 on Build GRCh38
rs777631196 Chr.1: 89009301-89009301 on Build GRCh38
rs745829250 Chr.1: 89009186-89009186 on Build GRCh38
rs767743790 Chr.1: 89009360-89009360 on Build GRCh38
rs752297910 Chr.1: 89009215-89009215 on Build GRCh38
rs891336747 Chr.1: 89009275-89009275 on Build GRCh38
rs996777882 Chr.1: 89009230-89009230 on Build GRCh38
rs1004458985 Chr.1: 89009294-89009294 on Build GRCh38
rs748015277 Chr.1: 89009354-89009355 on Build GRCh38
rs144631115 Chr.1: 89009233-89009233 on Build GRCh38
rs993242183 Chr.1: 89009268-89009268 on Build GRCh38
rs879902650 Chr.1: 89009380-89009380 on Build GRCh38
rs201419296 Chr.1: 89009374-89009374 on Build GRCh38
rs138557169 Chr.1: 89009336-89009336 on Build GRCh38
rs928829820 Chr.1: 89009363-89009363 on Build GRCh38
rs768756939 Chr.1: 89009189-89009189 on Build GRCh38
rs867572188 Chr.1: 89009194-89009194 on Build GRCh38
rs1007782985 Chr.1: 89009205-89009205 on Build GRCh38
rs774363421 Chr.1: 89009347-89009347 on Build GRCh38
rs367916921 Chr.1: 89009206-89009206 on Build GRCh38
rs773520369 Chr.1: 89009366-89009366 on Build GRCh38
rs141209926 Chr.1: 89009397-89009397 on Build GRCh38
rs773270062 Chr.1: 89009385-89009386 on Build GRCh38
rs200901183 Chr.1: 89009386-89009386 on Build GRCh38
rs761959766 Chr.1: 89009356-89009356 on Build GRCh38

View Full Product Details