Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113115443-113115698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F7
Entrez Gene ID
2155
Gene Name
coagulation factor VII
Gene Aliases
SPCA
UniGene
Hs.36989
Gene Chromosome Location
Chr.13: 113105773-113120681 on Build GRCh38
Transcripts
NM_000131.4 NM_001267554.1 NM_019616.3 NR_051961.1 XM_006719963.3 XM_011537474.2 XM_011537475.2 XM_011537476.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657775 c.258C>T p.N86N Chr.13: 113115694-113115694 on Build GRCh38
6554171 c.442C>T p.Q148* Chr.13: 113115671-113115671 on Build GRCh38
7422782 c.466G>A p.G156S Chr.13: 113115695-113115695 on Build GRCh38
5551627 c.232G>A p.D78N Chr.13: 113115668-113115668 on Build GRCh38
7422783 c.259G>A p.G87S Chr.13: 113115695-113115695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916477419 Chr.13: 113115480-113115480 on Build GRCh38
rs372514592 Chr.13: 113115612-113115612 on Build GRCh38
rs377588449 Chr.13: 113115613-113115613 on Build GRCh38
rs549926843 Chr.13: 113115570-113115570 on Build GRCh38
rs996064589 Chr.13: 113115588-113115588 on Build GRCh38
rs893671344 Chr.13: 113115481-113115481 on Build GRCh38
rs546722451 Chr.13: 113115471-113115471 on Build GRCh38
rs762144072 Chr.13: 113115610-113115610 on Build GRCh38
rs539382632 Chr.13: 113115504-113115504 on Build GRCh38
rs763636995 Chr.13: 113115635-113115635 on Build GRCh38
rs898602083 Chr.13: 113115584-113115584 on Build GRCh38
rs749421558 Chr.13: 113115505-113115505 on Build GRCh38
rs1019264714 Chr.13: 113115638-113115638 on Build GRCh38
rs767070264 Chr.13: 113115629-113115629 on Build GRCh38
rs890381946 Chr.13: 113115519-113115519 on Build GRCh38
rs569589966 Chr.13: 113115522-113115522 on Build GRCh38
rs752078675 Chr.13: 113115630-113115630 on Build GRCh38
rs763403619 Chr.13: 113115620-113115620 on Build GRCh38
rs143572954 Chr.13: 113115599-113115599 on Build GRCh38
rs753712213 Chr.13: 113115644-113115644 on Build GRCh38
rs536852622 Chr.13: 113115549-113115549 on Build GRCh38
rs569557 Chr.13: 113115603-113115603 on Build GRCh38
rs778809670 Chr.13: 113115658-113115658 on Build GRCh38
rs750407531 Chr.13: 113115666-113115666 on Build GRCh38
rs768984801 Chr.13: 113115530-113115530 on Build GRCh38
rs1014878469 Chr.13: 113115589-113115589 on Build GRCh38
rs190239932 Chr.13: 113115517-113115517 on Build GRCh38
rs757087734 Chr.13: 113115646-113115646 on Build GRCh38
rs760439688 Chr.13: 113115634-113115634 on Build GRCh38
rs72663561 Chr.13: 113115488-113115488 on Build GRCh38
rs940078804 Chr.13: 113115559-113115559 on Build GRCh38
rs773471899 Chr.13: 113115607-113115607 on Build GRCh38

View Full Product Details