Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 190099016-190099241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP3
Entrez Gene ID
339479
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
DBCCR1L, DBCCR1L1, FAM5C
UniGene
Hs.65765
Gene Chromosome Location
Chr.1: 190097662-190477997 on Build GRCh38
Transcripts
NM_001317188.1 NM_199051.2 XM_011509472.2 XM_011509475.2 XM_011509476.2 XM_017001125.1 XM_017001126.1 XM_017001127.1 XM_017001128.1 XM_017001129.1 XM_017001130.1 XM_017001131.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901276 c.1274C>T p.S425L Chr.1: 190099045-190099045 on Build GRCh38
6478743 c.1245C>T p.L415L Chr.1: 190099074-190099074 on Build GRCh38
6659899 c.1254C>G p.S418R Chr.1: 190099065-190099065 on Build GRCh38
7198527 c.1297G>A p.V433M Chr.1: 190099022-190099022 on Build GRCh38
4764200 c.1229G>A p.C410Y Chr.1: 190099090-190099090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376124518 Chr.1: 190099139-190099139 on Build GRCh38
rs552714085 Chr.1: 190099210-190099210 on Build GRCh38
rs748884368 Chr.1: 190099077-190099077 on Build GRCh38
rs34098782 Chr.1: 190099078-190099078 on Build GRCh38
rs757210693 Chr.1: 190099038-190099038 on Build GRCh38
rs370578801 Chr.1: 190099145-190099145 on Build GRCh38
rs1016373683 Chr.1: 190099199-190099199 on Build GRCh38
rs963448813 Chr.1: 190099211-190099211 on Build GRCh38
rs764002569 Chr.1: 190099135-190099135 on Build GRCh38
rs749022277 Chr.1: 190099175-190099175 on Build GRCh38
rs932101955 Chr.1: 190099125-190099125 on Build GRCh38
rs539008222 Chr.1: 190099198-190099198 on Build GRCh38
rs779404004 Chr.1: 190099173-190099173 on Build GRCh38
rs781215603 Chr.1: 190099155-190099155 on Build GRCh38
rs755712395 Chr.1: 190099162-190099162 on Build GRCh38
rs760751290 Chr.1: 190099113-190099113 on Build GRCh38
rs911953658 Chr.1: 190099160-190099160 on Build GRCh38
rs373147254 Chr.1: 190099137-190099137 on Build GRCh38
rs762332585 Chr.1: 190099104-190099104 on Build GRCh38
rs759653868 Chr.1: 190099134-190099134 on Build GRCh38
rs769108806 Chr.1: 190099079-190099079 on Build GRCh38
rs138103319 Chr.1: 190099161-190099161 on Build GRCh38
rs769342168 Chr.1: 190099071-190099071 on Build GRCh38
rs774680022 Chr.1: 190099093-190099093 on Build GRCh38
rs768148345 Chr.1: 190099184-190099184 on Build GRCh38
rs745308648 Chr.1: 190099051-190099051 on Build GRCh38
rs567209093 Chr.1: 190099187-190099187 on Build GRCh38
rs546624036 Chr.1: 190099114-190099114 on Build GRCh38
rs754129370 Chr.1: 190099130-190099130 on Build GRCh38
rs772531253 Chr.1: 190099112-190099112 on Build GRCh38
rs775267735 Chr.1: 190099075-190099075 on Build GRCh38
rs148343462 Chr.1: 190099085-190099085 on Build GRCh38
rs866895040 Chr.1: 190099055-190099055 on Build GRCh38
rs1036264238 Chr.1: 190099159-190099159 on Build GRCh38
rs780878224 Chr.1: 190099044-190099044 on Build GRCh38
rs555067902 Chr.1: 190099183-190099183 on Build GRCh38

View Full Product Details