Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36386074-36386343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
UniGene
Hs.75238
Gene Chromosome Location
Chr.21: 36380145-36416827 on Build GRCh38
Transcripts
NM_005441.2 XM_011529755.2 XM_017028477.1 XM_017028478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4618103 c.98C>T p.A33V Chr.21: 36386234-36386234 on Build GRCh38
6161348 c.112G>A p.D38N Chr.21: 36386248-36386248 on Build GRCh38
74133 c.112G>T p.D38Y Chr.21: 36386248-36386248 on Build GRCh38
4134918 c.54C>T p.S18S Chr.21: 36386190-36386190 on Build GRCh38
1637483 c.39G>T p.K13N Chr.21: 36386175-36386175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772699844 Chr.21: 36386309-36386309 on Build GRCh38
rs758301606 Chr.21: 36386273-36386273 on Build GRCh38
rs143543321 Chr.21: 36386183-36386183 on Build GRCh38
rs1042074120 Chr.21: 36386126-36386126 on Build GRCh38
rs372478630 Chr.21: 36386114-36386114 on Build GRCh38
rs771454096 Chr.21: 36386110-36386110 on Build GRCh38
rs772810675 Chr.21: 36386235-36386235 on Build GRCh38
rs767953331 Chr.21: 36386144-36386144 on Build GRCh38
rs776653217 Chr.21: 36386257-36386257 on Build GRCh38
rs373321906 Chr.21: 36386300-36386300 on Build GRCh38
rs549681857 Chr.21: 36386274-36386274 on Build GRCh38
rs754734966 Chr.21: 36386168-36386170 on Build GRCh38
rs755812719 Chr.21: 36386295-36386295 on Build GRCh38
rs764534792 Chr.21: 36386253-36386253 on Build GRCh38
rs749061160 Chr.21: 36386106-36386106 on Build GRCh38
rs748844043 Chr.21: 36386302-36386302 on Build GRCh38
rs758358794 Chr.21: 36386207-36386207 on Build GRCh38
rs760123435 Chr.21: 36386124-36386124 on Build GRCh38
rs780764051 Chr.21: 36386223-36386223 on Build GRCh38
rs757229798 Chr.21: 36386199-36386199 on Build GRCh38
rs539243544 Chr.21: 36386174-36386174 on Build GRCh38
rs1000665395 Chr.21: 36386190-36386190 on Build GRCh38
rs1033106706 Chr.21: 36386196-36386196 on Build GRCh38
rs761186456 Chr.21: 36386248-36386248 on Build GRCh38
rs776014948 Chr.21: 36386161-36386161 on Build GRCh38
rs778748692 Chr.21: 36386308-36386309 on Build GRCh38
rs536115732 Chr.21: 36386270-36386270 on Build GRCh38
rs376058259 Chr.21: 36386181-36386181 on Build GRCh38
rs889273654 Chr.21: 36386210-36386210 on Build GRCh38
rs373045673 Chr.21: 36386242-36386242 on Build GRCh38
rs747714663 Chr.21: 36386291-36386291 on Build GRCh38
rs781044532 Chr.21: 36386287-36386287 on Build GRCh38
rs765367431 Chr.21: 36386202-36386202 on Build GRCh38
rs762129577 Chr.21: 36386260-36386260 on Build GRCh38
rs369167961 Chr.21: 36386286-36386286 on Build GRCh38
rs972346102 Chr.21: 36386289-36386289 on Build GRCh38
rs765185475 Chr.21: 36386263-36386263 on Build GRCh38
rs747848592 Chr.21: 36386229-36386229 on Build GRCh38
rs761298518 Chr.21: 36386107-36386107 on Build GRCh38
rs369391435 Chr.21: 36386241-36386241 on Build GRCh38
rs750325153 Chr.21: 36386204-36386204 on Build GRCh38
rs779803781 Chr.21: 36386209-36386209 on Build GRCh38
rs375101149 Chr.21: 36386280-36386280 on Build GRCh38
rs142093091 Chr.21: 36386218-36386218 on Build GRCh38

View Full Product Details