Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49340060-49340321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
UniGene
Hs.166556
Gene Chromosome Location
Chr.19: 49335314-49340608 on Build GRCh38
Transcripts
NM_001040031.1 NM_001774.2 XM_005259435.3 XM_005259436.3 XM_011527542.2 XM_011527543.2 XM_011527544.2 XM_017027513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585425 c.769-6C>T p.? Chr.19: 49340245-49340245 on Build GRCh38
3536791 c.810G>T p.L270L Chr.19: 49340292-49340292 on Build GRCh38
1612572 c.828G>T p.R276R Chr.19: 49340310-49340310 on Build GRCh38
2752404 c.811G>C p.D271H Chr.19: 49340293-49340293 on Build GRCh38
6085773 c.831C>A p.L277L Chr.19: 49340313-49340313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754379575 Chr.19: 49340252-49340252 on Build GRCh38
rs1007186219 Chr.19: 49340114-49340114 on Build GRCh38
rs750066874 Chr.19: 49340167-49340167 on Build GRCh38
rs904481242 Chr.19: 49340094-49340094 on Build GRCh38
rs761266164 Chr.19: 49340228-49340228 on Build GRCh38
rs754185548 Chr.19: 49340150-49340150 on Build GRCh38
rs201538114 Chr.19: 49340174-49340174 on Build GRCh38
rs764564773 Chr.19: 49340141-49340141 on Build GRCh38
rs371455160 Chr.19: 49340217-49340217 on Build GRCh38
rs947668434 Chr.19: 49340082-49340082 on Build GRCh38
rs765604817 Chr.19: 49340268-49340268 on Build GRCh38
rs376634983 Chr.19: 49340241-49340241 on Build GRCh38
rs759775399 Chr.19: 49340211-49340211 on Build GRCh38
rs750073378 Chr.19: 49340279-49340279 on Build GRCh38
rs757688722 Chr.19: 49340156-49340156 on Build GRCh38
rs1021410456 Chr.19: 49340089-49340089 on Build GRCh38
rs896872099 Chr.19: 49340171-49340171 on Build GRCh38
rs1044636996 Chr.19: 49340103-49340103 on Build GRCh38
rs865836483 Chr.19: 49340187-49340187 on Build GRCh38
rs774489141 Chr.19: 49340207-49340207 on Build GRCh38
rs566279941 Chr.19: 49340243-49340243 on Build GRCh38
rs976206118 Chr.19: 49340178-49340178 on Build GRCh38
rs867888318 Chr.19: 49340219-49340219 on Build GRCh38
rs762317085 Chr.19: 49340253-49340253 on Build GRCh38
rs1056827837 Chr.19: 49340127-49340127 on Build GRCh38
rs554442277 Chr.19: 49340182-49340182 on Build GRCh38
rs760764774 Chr.19: 49340154-49340155 on Build GRCh38
rs776792516 Chr.19: 49340180-49340180 on Build GRCh38
rs1018178030 Chr.19: 49340245-49340245 on Build GRCh38
rs780971385 Chr.19: 49340183-49340183 on Build GRCh38
rs758018839 Chr.19: 49340292-49340292 on Build GRCh38
rs376872337 Chr.19: 49340198-49340198 on Build GRCh38
rs779445350 Chr.19: 49340298-49340298 on Build GRCh38
rs535519912 Chr.19: 49340169-49340169 on Build GRCh38
rs773450117 Chr.19: 49340125-49340125 on Build GRCh38
rs760123514 Chr.19: 49340223-49340224 on Build GRCh38
rs1010047075 Chr.19: 49340088-49340088 on Build GRCh38
rs756651146 Chr.19: 49340128-49340128 on Build GRCh38
rs746944507 Chr.19: 49340181-49340181 on Build GRCh38
rs766501876 Chr.19: 49340167-49340168 on Build GRCh38
rs909374728 Chr.19: 49340265-49340265 on Build GRCh38
rs765835577 Chr.19: 49340236-49340238 on Build GRCh38
rs753161503 Chr.19: 49340245-49340246 on Build GRCh38
rs775578713 Chr.19: 49340224-49340224 on Build GRCh38
rs748003108 Chr.19: 49340195-49340195 on Build GRCh38
rs112054750 Chr.19: 49340109-49340109 on Build GRCh38

View Full Product Details