Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177280260-177280533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP2
Entrez Gene ID
57795
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
DBCCR1L2, FAM5B
UniGene
Hs.495918
Gene Chromosome Location
Chr.1: 177170922-177282422 on Build GRCh38
Transcripts
NM_021165.3 XM_005245379.1 XM_011509826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6876806 c.1268C>T p.S423F Chr.1: 177280444-177280444 on Build GRCh38
5462093 c.1331C>T p.T444I Chr.1: 177280507-177280507 on Build GRCh38
4568728 c.1295T>C p.F432S Chr.1: 177280471-177280471 on Build GRCh38
4464802 c.1352C>T p.S451F Chr.1: 177280528-177280528 on Build GRCh38
2094333 c.1285G>A p.E429K Chr.1: 177280461-177280461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773076075 Chr.1: 177280451-177280451 on Build GRCh38
rs772103592 Chr.1: 177280356-177280356 on Build GRCh38
rs183482301 Chr.1: 177280336-177280336 on Build GRCh38
rs759785259 Chr.1: 177280500-177280500 on Build GRCh38
rs772053075 Chr.1: 177280449-177280449 on Build GRCh38
rs747115146 Chr.1: 177280464-177280464 on Build GRCh38
rs886984592 Chr.1: 177280340-177280340 on Build GRCh38
rs764439252 Chr.1: 177280410-177280410 on Build GRCh38
rs765607642 Chr.1: 177280394-177280394 on Build GRCh38
rs867989484 Chr.1: 177280299-177280299 on Build GRCh38
rs776789362 Chr.1: 177280377-177280377 on Build GRCh38
rs539302390 Chr.1: 177280432-177280432 on Build GRCh38
rs567769287 Chr.1: 177280321-177280321 on Build GRCh38
rs748101642 Chr.1: 177280448-177280448 on Build GRCh38
rs758937117 Chr.1: 177280407-177280407 on Build GRCh38
rs753196685 Chr.1: 177280396-177280396 on Build GRCh38
rs760682201 Chr.1: 177280364-177280364 on Build GRCh38
rs763167273 Chr.1: 177280507-177280507 on Build GRCh38
rs904557773 Chr.1: 177280468-177280468 on Build GRCh38
rs369846504 Chr.1: 177280433-177280433 on Build GRCh38
rs749962289 Chr.1: 177280412-177280412 on Build GRCh38
rs192059834 Chr.1: 177280327-177280327 on Build GRCh38
rs778053740 Chr.1: 177280346-177280346 on Build GRCh38
rs773339747 Chr.1: 177280358-177280358 on Build GRCh38
rs766603057 Chr.1: 177280365-177280365 on Build GRCh38
rs1004434154 Chr.1: 177280399-177280399 on Build GRCh38
rs1056301601 Chr.1: 177280335-177280336 on Build GRCh38
rs188772885 Chr.1: 177280323-177280323 on Build GRCh38
rs759929937 Chr.1: 177280383-177280383 on Build GRCh38
rs996183020 Chr.1: 177280501-177280501 on Build GRCh38
rs866141858 Chr.1: 177280487-177280487 on Build GRCh38
rs758693131 Chr.1: 177280326-177280326 on Build GRCh38
rs752385924 Chr.1: 177280311-177280311 on Build GRCh38
rs905137362 Chr.1: 177280322-177280322 on Build GRCh38
rs776882239 Chr.1: 177280498-177280498 on Build GRCh38
rs775924092 Chr.1: 177280506-177280506 on Build GRCh38
rs778753983 Chr.1: 177280435-177280435 on Build GRCh38
rs765580916 Chr.1: 177280502-177280502 on Build GRCh38
rs770962414 Chr.1: 177280496-177280496 on Build GRCh38
rs906108217 Chr.1: 177280319-177280319 on Build GRCh38

View Full Product Details