Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155298642-155298897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC2
Entrez Gene ID
1193
Gene Name
chloride intracellular channel 2
Gene Aliases
CLIC2b, MRXS32, XAP121
UniGene
Hs.655445
Gene Chromosome Location
Chr.X: 155276207-155334681 on Build GRCh38
Transcripts
NM_001289.5

SNP Detail

SNP ID Chromosome Location
rs781838316 Chr.X: 155298794-155298794 on Build GRCh38
rs782810557 Chr.X: 155298767-155298767 on Build GRCh38
rs1033024813 Chr.X: 155298688-155298688 on Build GRCh38
rs191627909 Chr.X: 155298814-155298814 on Build GRCh38
rs782270791 Chr.X: 155298866-155298866 on Build GRCh38
rs200903076 Chr.X: 155298731-155298731 on Build GRCh38
rs781853876 Chr.X: 155298817-155298817 on Build GRCh38
rs967604546 Chr.X: 155298673-155298673 on Build GRCh38
rs782667135 Chr.X: 155298837-155298837 on Build GRCh38
rs372701786 Chr.X: 155298752-155298752 on Build GRCh38
rs201668560 Chr.X: 155298858-155298858 on Build GRCh38
rs782548834 Chr.X: 155298819-155298819 on Build GRCh38
rs782349599 Chr.X: 155298748-155298748 on Build GRCh38
rs376246918 Chr.X: 155298871-155298871 on Build GRCh38
rs781849311 Chr.X: 155298765-155298765 on Build GRCh38
rs782688201 Chr.X: 155298785-155298785 on Build GRCh38
rs782170553 Chr.X: 155298679-155298679 on Build GRCh38
rs782081314 Chr.X: 155298799-155298799 on Build GRCh38
rs782650478 Chr.X: 155298749-155298749 on Build GRCh38
rs781800421 Chr.X: 155298839-155298839 on Build GRCh38
rs782067711 Chr.X: 155298780-155298780 on Build GRCh38
rs183067063 Chr.X: 155298865-155298865 on Build GRCh38

View Full Product Details