Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240630511-240630758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR35
Entrez Gene ID
2859
Gene Name
G protein-coupled receptor 35
Gene Aliases
-
UniGene
Hs.112218
Gene Chromosome Location
Chr.2: 240605408-240631259 on Build GRCh38
Transcripts
NM_001195381.1 NM_001195382.1 NM_005301.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3579931 c.783C>A p.T261T Chr.2: 240630735-240630735 on Build GRCh38
3579929 c.602G>A p.R201K Chr.2: 240630554-240630554 on Build GRCh38
5396621 c.613G>A p.D205N Chr.2: 240630565-240630565 on Build GRCh38
5396620 c.706G>A p.D236N Chr.2: 240630565-240630565 on Build GRCh38
5002595 c.622C>A p.Q208K Chr.2: 240630574-240630574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369527208 Chr.2: 240630534-240630534 on Build GRCh38
rs779192596 Chr.2: 240630723-240630723 on Build GRCh38
rs764395578 Chr.2: 240630553-240630553 on Build GRCh38
rs752386747 Chr.2: 240630663-240630663 on Build GRCh38
rs763024464 Chr.2: 240630544-240630544 on Build GRCh38
rs201441371 Chr.2: 240630568-240630568 on Build GRCh38
rs530284304 Chr.2: 240630564-240630564 on Build GRCh38
rs548051621 Chr.2: 240630536-240630536 on Build GRCh38
rs139042901 Chr.2: 240630707-240630707 on Build GRCh38
rs138617727 Chr.2: 240630690-240630690 on Build GRCh38
rs773286501 Chr.2: 240630636-240630636 on Build GRCh38
rs199886367 Chr.2: 240630650-240630650 on Build GRCh38
rs543091271 Chr.2: 240630721-240630721 on Build GRCh38
rs12468453 Chr.2: 240630675-240630675 on Build GRCh38
rs140850565 Chr.2: 240630602-240630602 on Build GRCh38
rs559709613 Chr.2: 240630562-240630562 on Build GRCh38
rs772297163 Chr.2: 240630685-240630685 on Build GRCh38
rs770344320 Chr.2: 240630601-240630601 on Build GRCh38
rs900052891 Chr.2: 240630596-240630596 on Build GRCh38
rs373558269 Chr.2: 240630655-240630655 on Build GRCh38
rs763867971 Chr.2: 240630715-240630715 on Build GRCh38
rs776612723 Chr.2: 240630704-240630704 on Build GRCh38
rs145963376 Chr.2: 240630676-240630676 on Build GRCh38
rs779033520 Chr.2: 240630568-240630570 on Build GRCh38
rs890489173 Chr.2: 240630734-240630734 on Build GRCh38
rs761849391 Chr.2: 240630561-240630561 on Build GRCh38
rs538583396 Chr.2: 240630647-240630647 on Build GRCh38
rs987404469 Chr.2: 240630608-240630608 on Build GRCh38
rs762078393 Chr.2: 240630631-240630631 on Build GRCh38
rs142716446 Chr.2: 240630589-240630589 on Build GRCh38
rs768941519 Chr.2: 240630623-240630623 on Build GRCh38
rs963493042 Chr.2: 240630586-240630586 on Build GRCh38
rs754538522 Chr.2: 240630571-240630571 on Build GRCh38
rs147827675 Chr.2: 240630711-240630711 on Build GRCh38
rs766022798 Chr.2: 240630567-240630567 on Build GRCh38
rs147700491 Chr.2: 240630618-240630618 on Build GRCh38
rs753537893 Chr.2: 240630654-240630654 on Build GRCh38
rs756768919 Chr.2: 240630718-240630718 on Build GRCh38
rs760776139 Chr.2: 240630697-240630697 on Build GRCh38
rs758193886 Chr.2: 240630665-240630665 on Build GRCh38
rs766362917 Chr.2: 240630652-240630652 on Build GRCh38
rs554457295 Chr.2: 240630719-240630719 on Build GRCh38
rs746538122 Chr.2: 240630667-240630667 on Build GRCh38
rs770957036 Chr.2: 240630699-240630699 on Build GRCh38
rs778659810 Chr.2: 240630584-240630584 on Build GRCh38
rs12468485 Chr.2: 240630710-240630710 on Build GRCh38
rs370613558 Chr.2: 240630671-240630671 on Build GRCh38
rs756109000 Chr.2: 240630565-240630565 on Build GRCh38
rs1057346816 Chr.2: 240630678-240630678 on Build GRCh38
rs575778583 Chr.2: 240630720-240630720 on Build GRCh38
rs767578418 Chr.2: 240630633-240630633 on Build GRCh38
rs775924140 Chr.2: 240630541-240630541 on Build GRCh38
rs746040312 Chr.2: 240630714-240630714 on Build GRCh38
rs376362528 Chr.2: 240630590-240630590 on Build GRCh38
rs144191361 Chr.2: 240630708-240630708 on Build GRCh38
rs746376617 Chr.2: 240630595-240630595 on Build GRCh38
rs558162538 Chr.2: 240630730-240630730 on Build GRCh38
rs751293317 Chr.2: 240630716-240630716 on Build GRCh38
rs777357729 Chr.2: 240630666-240630666 on Build GRCh38
rs76041925 Chr.2: 240630630-240630630 on Build GRCh38
rs377190882 Chr.2: 240630670-240630670 on Build GRCh38
rs777486037 Chr.2: 240630591-240630591 on Build GRCh38

View Full Product Details