Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74926693-74926946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX5A
Entrez Gene ID
9377
Gene Name
cytochrome c oxidase subunit 5A
Gene Aliases
COX, COX-VA, VA
UniGene
Hs.401903
Gene Chromosome Location
Chr.15: 74920275-74938154 on Build GRCh38
Transcripts
NM_004255.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427525 c.231T>A p.L77L Chr.15: 74926874-74926874 on Build GRCh38
6372552 c.301G>T p.D101Y Chr.15: 74926804-74926804 on Build GRCh38
1749312 c.241G>A p.D81N Chr.15: 74926864-74926864 on Build GRCh38
1938964 c.319C>T p.R107C Chr.15: 74926786-74926786 on Build GRCh38
7078523 c.274G>A p.A92T Chr.15: 74926831-74926831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200875315 Chr.15: 74926849-74926849 on Build GRCh38
rs764213271 Chr.15: 74926919-74926919 on Build GRCh38
rs148865910 Chr.15: 74926811-74926811 on Build GRCh38
rs752698153 Chr.15: 74926802-74926802 on Build GRCh38
rs765056380 Chr.15: 74926885-74926885 on Build GRCh38
rs557275991 Chr.15: 74926867-74926867 on Build GRCh38
rs746397506 Chr.15: 74926825-74926825 on Build GRCh38
rs767044165 Chr.15: 74926726-74926726 on Build GRCh38
rs760804502 Chr.15: 74926755-74926756 on Build GRCh38
rs546553770 Chr.15: 74926747-74926747 on Build GRCh38
rs151002462 Chr.15: 74926877-74926877 on Build GRCh38
rs778137192 Chr.15: 74926812-74926812 on Build GRCh38
rs4131311 Chr.15: 74926766-74926766 on Build GRCh38
rs377436159 Chr.15: 74926909-74926909 on Build GRCh38
rs747437553 Chr.15: 74926813-74926813 on Build GRCh38
rs199907556 Chr.15: 74926855-74926855 on Build GRCh38
rs760257324 Chr.15: 74926872-74926872 on Build GRCh38
rs773053707 Chr.15: 74926734-74926734 on Build GRCh38
rs773949037 Chr.15: 74926835-74926835 on Build GRCh38
rs140891101 Chr.15: 74926878-74926878 on Build GRCh38
rs542455445 Chr.15: 74926853-74926853 on Build GRCh38
rs759571153 Chr.15: 74926883-74926883 on Build GRCh38
rs765384044 Chr.15: 74926790-74926790 on Build GRCh38
rs757699339 Chr.15: 74926816-74926816 on Build GRCh38
rs976996622 Chr.15: 74926723-74926723 on Build GRCh38
rs142618460 Chr.15: 74926839-74926839 on Build GRCh38
rs751861358 Chr.15: 74926921-74926921 on Build GRCh38
rs764263218 Chr.15: 74926892-74926892 on Build GRCh38
rs754834730 Chr.15: 74926789-74926789 on Build GRCh38
rs1800994 Chr.15: 74926808-74926814 on Build GRCh38
rs368515990 Chr.15: 74926784-74926784 on Build GRCh38
rs781655016 Chr.15: 74926822-74926822 on Build GRCh38
rs752890652 Chr.15: 74926886-74926886 on Build GRCh38
rs754067907 Chr.15: 74926901-74926903 on Build GRCh38
rs766107845 Chr.15: 74926783-74926783 on Build GRCh38
rs770178135 Chr.15: 74926834-74926834 on Build GRCh38

View Full Product Details