Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123688840-123689065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA13
Entrez Gene ID
312
Gene Name
annexin A13
Gene Aliases
ANX13, ISA
UniGene
Hs.181107
Gene Chromosome Location
Chr.8: 123680794-123737426 on Build GRCh38
Transcripts
NM_001003954.2 NM_004306.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7118020 c.778G>C p.D260H Chr.8: 123688934-123688934 on Build GRCh38
7276493 c.810A>G p.S270S Chr.8: 123688902-123688902 on Build GRCh38
6305396 c.774C>A p.G258G Chr.8: 123688938-123688938 on Build GRCh38
3785864 c.807A>G p.T269T Chr.8: 123688905-123688905 on Build GRCh38
6457199 c.841G>A p.V281M Chr.8: 123688871-123688871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775205345 Chr.8: 123688920-123688920 on Build GRCh38
rs367655009 Chr.8: 123688872-123688872 on Build GRCh38
rs140025435 Chr.8: 123688917-123688917 on Build GRCh38
rs779382326 Chr.8: 123688974-123688974 on Build GRCh38
rs765213376 Chr.8: 123688919-123688919 on Build GRCh38
rs118049232 Chr.8: 123688957-123688957 on Build GRCh38
rs755318427 Chr.8: 123688970-123688970 on Build GRCh38
rs371442967 Chr.8: 123688883-123688883 on Build GRCh38
rs189031746 Chr.8: 123689002-123689002 on Build GRCh38
rs542478644 Chr.8: 123689007-123689007 on Build GRCh38
rs150006563 Chr.8: 123688896-123688896 on Build GRCh38
rs201252513 Chr.8: 123688895-123688895 on Build GRCh38
rs144282673 Chr.8: 123688898-123688898 on Build GRCh38
rs997919020 Chr.8: 123688900-123688900 on Build GRCh38
rs1020896941 Chr.8: 123689009-123689009 on Build GRCh38
rs969038897 Chr.8: 123689001-123689001 on Build GRCh38
rs771567427 Chr.8: 123688929-123688930 on Build GRCh38
rs34594422 Chr.8: 123688899-123688899 on Build GRCh38
rs770490089 Chr.8: 123688884-123688884 on Build GRCh38
rs55688638 Chr.8: 123688953-123688953 on Build GRCh38
rs981979614 Chr.8: 123689006-123689006 on Build GRCh38
rs757542864 Chr.8: 123688950-123688950 on Build GRCh38
rs751588826 Chr.8: 123688871-123688871 on Build GRCh38
rs376117970 Chr.8: 123688892-123688892 on Build GRCh38
rs756632548 Chr.8: 123688954-123688954 on Build GRCh38
rs890621608 Chr.8: 123688927-123688927 on Build GRCh38
rs749627143 Chr.8: 123688964-123688964 on Build GRCh38
rs772645900 Chr.8: 123688935-123688935 on Build GRCh38
rs927907756 Chr.8: 123689027-123689027 on Build GRCh38
rs746609568 Chr.8: 123688978-123688978 on Build GRCh38
rs781474899 Chr.8: 123688882-123688882 on Build GRCh38
rs751712187 Chr.8: 123688944-123688944 on Build GRCh38
rs764330501 Chr.8: 123688940-123688940 on Build GRCh38
rs1011295139 Chr.8: 123688994-123688994 on Build GRCh38
rs780553268 Chr.8: 123688885-123688885 on Build GRCh38
rs762657529 Chr.8: 123688930-123688930 on Build GRCh38

View Full Product Details